Skip to Content
Merck

1.41352

SAFC

Polyvinyl alcohol

26-88, EMPROVE® ESSENTIAL, Ph. Eur., ChP, USP, JPE

Pharma Manufacturing

Synonym(s):

Poly(vinyl alcohol), PVA, PVOH

Sign Into View Organizational & Contract Pricing

Select a Size

1 KG
€309.00
25 KG
Inquire

€309.00


Estimated to ship on24 May 2025


Request a Bulk OrderRequest a Sample

Select a Size

Change View
1 KG
€309.00
25 KG
Inquire

About This Item

Linear Formula:
[-CH2CHOH-]n
CAS Number:
MDL number:
UNSPSC Code:
13111042

€309.00


Estimated to ship on24 May 2025


Request a Bulk OrderRequest a Sample

Agency

ChP
JPE
Ph. Eur.
USP

Quality Level

description

EMPROVE® ESSENTIAL Ph Eur,USP,JPE

product line

EMPROVE® ESSENTIAL

form

scales

technique(s)

API processing | nano-milling: suitable

mp

160-240 °C

application(s)

controlled release
liquid formulation
ophthalmics
pharmaceutical
semi-solid formulation
solid formulation

storage temp.

15-25°C

InChI

1S/C2H4O/c1-2-3/h2-3H,1H2

InChI key

IMROMDMJAWUWLK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

When working on a pharmaceutical formulation, there are specific considerations to make and challenging hurdles to take before you can successfully launch your final drug product - formulation stability, release kinetics and bioavailability limitations just to name a few. With our application know-how and regulatory expertise, we support you in every step of development, scale-up, and production.
As part of our Emprove® Program, our raw materials are offered with extensive documentation facilitating compliance of your pharma and biopharma product, full supply chain transparency and risk mitigation. Our SAFC® portfolio of high-quality products for biopharmaceutical and pharmaceutical formulation and production withstands strict quality control procedures and is produced according to applicable cGMP guidelines.

Application

Polyvinyl alcohol 26-88 is a synthetic, bio-compatible, toxicologically safe, and multi-compendial polymer. It is part of a portfolio spanning a wide range of pharmaceutical grade polyvinyl alcohols of different viscosities, hydrolysis grades and molecular weights and is well suited for a broad variety of pharmaceutical applications, including solid, liquid, and semi-solid formulations.

Legal Information

Emprove is a registered trademark of Merck KGaA, Darmstadt, Germany
SAFC is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

What is the Emprove® Program?

The Emprove® Program is a system providing comprehensive and thorough documentation of our filters and single-use components, pharma raw materials, and starting materials. Four document types are included with an Emprove® Program subscription:

Material Qualification Dossier

Free of charge - Sign in to download

Quality Management Dossier (Open Access)

Free of charge - Sign in to download

Operational Excellence Dossier

Available by subscription or for a fee

Learn more about the benefits an Emprove® Program subscription can provide
Why are these documents unavailable?
Emprove® Dossiers contain information intended to support the registration process and risk assessment activities of pharmaceutical producers and contain confidential information. Please contact us if you would like access.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

PVA meets all the essential requirements to be used in ophthalmic preparations and offers important benefits compared to HPMC and CMC.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service