Skip to Content
Merck

1.06445

SAFC

Sodium L-glutamate monohydrate

EMPROVE® EXPERT, ChP, FCC, NF, E 621

Pharma Manufacturing

Synonym(s):

Sodium L-glutamate monohydrate, Sodium L-glutamate Monohydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H8NNaO4 · xH2O
CAS Number:
Molecular Weight:
169.11 (anhydrous basis)
MDL number:
UNSPSC Code:
12352209
E Number:
E621
EC Index Number:
205-538-1
Pricing and availability is not currently available.

Agency

ChP
NF

Quality Level

reg. compliance

FCC

product line

EMPROVE® EXPERT

form

solid

pH

6.7-7.2 (20 °C, 50 g/L in H2O)

mp

>200 °C (sublimed)

transition temp

flash point >900 °C

application(s)

liquid formulation
pharmaceutical
solid formulation

storage temp.

2-30°C

SMILES string

[Na+].N[C@@H](CCC(=O)[O-])C(=O)[O-].O.[H+]

InChI

1S/C5H9NO4.Na.H2O/c6-3(5(9)10)1-2-4(7)8;;/h3H,1-2,6H2,(H,7,8)(H,9,10);;1H2/q;+1;/p-1/t3-;;/m0../s1

InChI key

GJBHGUUFMNITCI-QTNFYWBSSA-M

General description

Finding the right excipient that matches your needs as well as regulatory demands can be a complicated challenge in formulation. With our application know-how and regulatory expertise, we support you in every step of development, scale-up, and production.
As part of our Emprove® Program, our raw materials are offered with extensive documentation facilitating compliance of your pharma and biopharma product, full supply chain transparency and risk mitigation. Our SAFC® portfolio of high-quality products for biopharmaceutical and pharmaceutical formulation and production withstands strict quality control procedures and is produced according to applicable cGMP guidelines.

Application

Sodium L-glutamate is commonly used as an ingredient in food and pharmaceutics. In pharmaceutical applications, sodium L-glutamate has been used primarily for oral dosage forms.

Legal Information

Emprove is a registered trademark of Merck KGaA, Darmstadt, Germany
SAFC is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


What is the Emprove® Program?

The Emprove® Program is a system providing comprehensive and thorough documentation of our filters and single-use components, pharma raw materials, and starting materials. Four document types are included with an Emprove® Program subscription:

Material Qualification Dossier

Free of charge - Sign in to download

Quality Management Dossier (Open Access)

Free of charge - Sign in to download

Operational Excellence Dossier

Available by subscription or for a fee

Learn more about the benefits an Emprove® Program subscription can provide
Why are these documents unavailable?
Emprove® Dossiers contain information intended to support the registration process and risk assessment activities of pharmaceutical producers and contain confidential information. Please contact us if you would like access.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service