Skip to Content
Merck

1.37045

SAFC

Lactose monohydrate

powder, EMPROVE® EXPERT, Ph. Eur., BP, ChP, NF, JP

Pharma Manufacturing

Synonym(s):

D-Lactose, β-D-Gal-(1→4)-D-Glc, 4-O-β-D-Galactopyranosyl-D-glucose, Milk sugar

Sign Into View Organizational & Contract Pricing

Select a Size

5 KG
€187.00
25 KG
€598.00

€187.00


Estimated to ship on01 June 2025


Request a Sample

Select a Size

Change View
5 KG
€187.00
25 KG
€598.00

About This Item

Empirical Formula (Hill Notation):
C12H22O11 · H2O
CAS Number:
Molecular Weight:
360.31
Beilstein:
3768231
MDL number:
UNSPSC Code:
12352201

€187.00


Estimated to ship on01 June 2025


Request a Sample

Agency

BP
ChP
JP
NF
Ph. Eur.

Quality Level

form

powder

manufacturer/tradename

EMPROVE® EXPERT

pH

4.0-6.5 (20 °C, 100 g/L in water)

mp

202 °C

solubility

161 g/L

density

500 kg/m3

suitability

passes test for IR spectroscopy
passes test for TLC
suitable for appearance

SMILES string

O.OC[C@@H](O)[C@@H](O[C@@H]1O[C@H](CO)[C@H](O)[C@H](O)[C@H]1O)[C@H](O)[C@@H](O)C=O

InChI

1S/C12H22O11.H2O/c13-1-4(16)7(18)11(5(17)2-14)23-12-10(21)9(20)8(19)6(3-15)22-12;/h1,4-12,14-21H,2-3H2;1H2/t4-,5+,6+,7+,8-,9-,10+,11+,12-;/m0./s1

InChI key

HBDJFVFTHLOSDW-XBLONOLSSA-N

Related Categories

Legal Information

Emprove is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


What is the Emprove® Program?

The Emprove® Program is a system providing comprehensive and thorough documentation of our filters and single-use components, pharma raw materials, and starting materials. Four document types are included with an Emprove® Program subscription:

Material Qualification Dossier

Free of charge - Sign in to download

Quality Management Dossier (Open Access)

Free of charge - Sign in to download

Operational Excellence Dossier

Available by subscription or for a fee

Learn more about the benefits an Emprove® Program subscription can provide
Why are these documents unavailable?
Emprove® Dossiers contain information intended to support the registration process and risk assessment activities of pharmaceutical producers and contain confidential information. Please contact us if you would like access.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service