Skip to Content
Merck
All Photos(1)

Key Documents

1.07660

Millipore

Lactose monohydrate

for biochemistry

Synonym(s):

Lactose monohydrate, Milk sugar, Lactobiose

Sign Into View Organizational & Contract Pricing

Select a Size

250 G
€60.20
1 KG
€138.00

€60.20


Estimated to ship on28 May 2025


Request a Bulk Order

Select a Size

Change View
250 G
€60.20
1 KG
€138.00

About This Item

Empirical Formula (Hill Notation):
C12H22O11 · xH2O
CAS Number:
Molecular Weight:
342.30 (anhydrous basis)
MDL number:
UNSPSC Code:
12352201
EC Index Number:
200-559-2
NACRES:
NA.79

€60.20


Estimated to ship on28 May 2025


Request a Bulk Order

Quality Level

form

powder

autoignition temp.

390 °C

potency

>10000 mg/kg LD50, oral (Rat)

pH

4.0-6.5 (20 °C, 100 g/L in H2O)

mp

202 °C

solubility

161 g/L

bulk density

500 kg/m3

storage temp.

15-25°C

SMILES string

O1[C@H]([C@@H]([C@H]([C@H]([C@H]1CO)O)O)O)O[C@@H]2[C@H](O[C@@H]([C@@H]([C@H]2O)O)O)CO

InChI

1S/C12H22O11/c13-1-3-5(15)6(16)9(19)12(22-3)23-10-4(2-14)21-11(20)8(18)7(10)17/h3-20H,1-2H2/t3-,4-,5+,6+,7-,8-,9-,10-,11+,12+/m1/s1

InChI key

GUBGYTABKSRVRQ-XLOQQCSPSA-N

Application


  • Insights into the potential of rheological measurements in development of dry powder inhalation formulations.: Explores the role of lactose monohydrate in the rheological properties of dry powder formulations, emphasizing its significance in developing more efficient and effective inhalable drugs (Almansour et al., 2022 Feb). Link to the article.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service