1.32406
HY-LiTE® 2 External Thermal Printer
For HY-LiTE® 2 (1.30100.0001) Instruments
Synonym(s):
External Printer, Printer, Strip printer
Select a Size
About This Item
Recommended Products
Agency
(HACCP, IATA)
ASTM®
application(s)
food and beverages
industrial hygiene
jet fuel
General description
Use of this optional strip printer allows for printing stored measurement results as well as auto-printing of results upon measurement completion.
The printer connects to the HY-LiTE® 2 instrument via included USB cable and can operate on either AC mains or battery power for flexible operation.
Kit Contents:
Thermal strip printer
Lithium-ion rechargeable battery (7.4V, 1500 mAh, 11.1 Wh)
- Roll thermal paper
Power supply unit (PSU)
PSU worldwide adapters (UK, EU, NA, AU)
USB cable
USB drive with product documentation
Printed QC certificate
For additional thermal roll paper, order item 1.30110.0002 Paper Roll Pack for HY-LiTE® 2 External Thermal Printer (12 rolls/pk).
Thermal paper roll specifications: 57mm direct thermal non-adhesive paper, 30mm* OD, 12.7mm* core. (*Tolerance: ±10%)
Application
Legal Information
Storage Class Code
11 - Combustible Solids
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service