Skip to Content
Merck
All Photos(7)

Documents

1.32372

Millipore

MC-Media Pad®

selective for bacteria, Detection of Enterobacteriaceae, pkg of 4 × 25 pieces

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116131
NACRES:
NA.75

sterility

sterile; aseptically processed

product line

MC-Media Pad®

form

liquid

feature

ready-to-use

packaging

pkg of 4 × 25 pieces

technique(s)

microbiological culture: suitable

application(s)

food and beverages
personnel monitoring
surface monitoring
water monitoring

storage temp.

2-8°C

suitability

selective for bacteria (Enterobacteriaceae)

General description

MC-MediaPad® is designed to ensure hygiene control in the food and beverage industry. Itsimplifies the process of detecting and enumerating Enterobacteriaceae, acommon quality indicator of food contamination. The MC-MediaPad® is a pre-sterilized, ready-to-use dry culture device, minimizing waste andsimplifying testing procedures. It contains a unique adhesive sheet, a test padcoated with medium and water absorption polymer, and a transparent cover film. The selective medium,pH indicator, and redox indicator enable specific and reliable detection. Uponinoculation, the sample diffuses through the pad via capillary action,automatically reconstituting the medium. The distinctive color change frompurple to red coloured colonies with a yellow zone in the test pad provides aclear visual indication of the presence of Enterobacteriaceae spp. presence.

Application

MC-Media Pad® is used for the enumeration andselective detection of Enterobacteriaceae spp in food samples from the food andbeverage industry.

Analysis Note

Count the red-coloured colonies with a yellow zone only. Colonies without the yellow zone should be ignored. In case of any difficulty in identifying the yellow zone, kindly count under a bright desk lamp or in a brighter environment.

Legal Information

MC MEDIA PAD is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service