Skip to Content
Merck
All Photos(1)

Key Documents

124033

Sigma-Aldrich

Akt Inhibitor XVI, AT7867

Synonym(s):

Akt Inhibitor XVI, AT7867, p70S6K Inhibitor III, 4-(4-Chlorophenyl)-4-(4-(1H-pyrazol-4-yl)-phenyl)-piperidine, PKA Inhibitor XIV, S6K1 Inhibitor III, Chlorophenyl-pyrazolyl-phenyl-piperidine

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
€232.00

€232.00


Estimated to ship on21 May 2025



Select a Size

Change View
10 MG
€232.00

About This Item

Empirical Formula (Hill Notation):
C20H20ClN3
Molecular Weight:
337.85
UNSPSC Code:
12352200
NACRES:
NA.77

€232.00


Estimated to ship on21 May 2025


Assay

≥99% (HPLC)

Quality Level

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

DMSO: 100 mg/mL

shipped in

wet ice

storage temp.

2-8°C

General description

A cell-permeable pyrazolo compound that acts as a potent inhibitor against Akt (IC50 = 32, 17, and 47 nM against Akt1, Akt2, and Akt3, respectively), PKA (IC50 = 20 nM), and p70S6K (IC50 = 85 nM) in an ATP-competitive manner (Ki = 18 nM using AKT2), with much reduced activity toward RSK1 (IC50 >100 nM) and a panel of 13 other cellular kinases (IC50 >1 µM), including PDK1, PI 3-Kβ, SGK, GSK-3β, and MEK1. Shown to effectively reduce the phosphorylation levels of GSK-3β, S6R, and FKHRL1 (IC50 range from 2.08 to 4.45 µM against GSK-3β pSer9 level), as well as inhibit the proliferation (IC50 range from 0.94 to 11.86 µM) of cancer cells exhibiting constitutive Akt pathway activation, including the PTEN-deficient MES-SA uterine sarcoma and U87MG glioblastoma cell lines. AT7867 treatments via i.p. or p.o. (20 mg/kg and 90 mg/kg, respectively, once every 3 days) are also reported to result in Akt pathway and tumor growth inhibition in mice bearing MES-SA (~40% of untreated control on day 10 by i.p. or p.o.) or U87MG (~50% of untreated control on day 10 by i.p.) xenograft in vivo.

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Grimshaw, K.M., et al. 2010. Mol. Cancer Ther.9, 1100.
Saxty, G., et al. 2007. J. Med. Chem.50, 2293.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service