Skip to Content
Merck
All Photos(1)

Key Documents

SML2194

Sigma-Aldrich

SNS-314 mesylate

≥98% (HPLC)

Synonym(s):

1-(3-Chlorophenyl)-3-(5-(2-(thieno[3,2-d]pyrimidin-4-ylamino)ethyl)thiazol-2-yl)urea methanesulfonate, N-(3-Chlorophenyl)-N′-[5-[2-(thieno[3,2-d]pyrimidin-4-ylamino)ethyl]-2-thiazolyl]urea methanesulfonate, SNS 314, SNS 314 mesylate, SNS314, SNS314 mesylate

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€55.80
25 MG
€225.00

€55.80


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
€55.80
25 MG
€225.00

About This Item

Empirical Formula (Hill Notation):
C18H15ClN6OS2 · CH4O3S
CAS Number:
Molecular Weight:
527.04
MDL number:
UNSPSC Code:
12352200

€55.80


Check Cart for Availability

Request a Bulk Order

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated
under inert gas

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

−20°C

SMILES string

[S](=O)(=O)(O)C.[s]1c(ncc1CCNc3ncnc4c3[s]cc4)NC(=O)Nc2cc(ccc2)Cl

InChI

1S/C18H15ClN6OS2.CH4O3S/c19-11-2-1-3-12(8-11)24-17(26)25-18-21-9-13(28-18)4-6-20-16-15-14(5-7-27-15)22-10-23-16;1-5(2,3)4/h1-3,5,7-10H,4,6H2,(H,20,22,23)(H2,21,24,25,26);1H3,(H,2,3,4)

InChI key

FYCODPVDEFFWSR-UHFFFAOYSA-N

Biochem/physiol Actions

Potent and selective pan-aurora kinase inhibitor that binds aurora kinase in the DFG-in conformation and exhibits anticancer efficacy in vitro and in vivo.
SNS-314 is a potent pan-aurora kinases inhibitor (IC50 = 9/31/3 nM against aurora kinase A/B/C or aurora 2/1/3) that binds aurora kinase in the DFG-in conformation and exhibits great selectivity against a panel of 219 kinases (>5-, >12-, >14-, >15-, >82-, >84-fold selectivity for aurora A over TrkB, TrkA, Flt4, Fms, DDR2, Axl, respectively; >100-fold selectivity over c-Raf and the remaining kinases). SNS-314 downregulates cellular histone H3 Ser10 (HH3 Ser10) phosphorylation (IC50 <16 nM) and exhibits potent antiproliferation activity in HCT116 human colon cancer cultures in vitro (IC50 = 5 nM). When applied in vivo, HCT116 likewise reduces pHH3 (Ser10) level in tumor tissue (by 75-100% 6 hrs post 50 mg/kg i.p. dosage) and suppresses tumor growth (100 mg/kg/day, 5 days on, 9 days off; 150 mg/kg biweekly x3), as well as potentiates docetaxel antitumor efficacy in a mouse HCT116 xenograft model in vivo (42.5 mg SNS-314/kg followed by 10 mg docetaxel/kg 24 hr later; biweekly x3).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jennifer P Arbitrario et al.
Cancer chemotherapy and pharmacology, 65(4), 707-717 (2009-08-04)
The Aurora family of serine/threonine kinases (Aurora-A, Aurora-B, and Aurora-C) plays a key role in cells orderly progression through mitosis. Elevated expression levels of Aurora kinases have been detected in a high percentage of melanoma, colon, breast, ovarian, gastric, and
Vincenzo Quagliariello et al.
Journal of cellular physiology, 231(8), 1784-1795 (2015-12-15)
The aim of this paper is based on the use of a hyaluronic acid hydrogel of Quercetin tested alone and in combination to an inhibitor of Aurora Kinase type A and B (SNS-314) on human medullary and papillary thyroid cancer
Antonella Antignani et al.
PloS one, 11(8), e0161415-e0161415 (2016-08-25)
The intersection of small molecular weight drugs and antibody-based therapeutics is rarely studied in large scale. Both types of agents are currently part of the cancer armamentarium. However, very little is known about how to combine them in optimal ways.
Raffaele Pezzani et al.
Investigational new drugs, 34(5), 531-540 (2016-05-15)
New therapeutic targets are needed to fight cancer. Aurora kinases (AK) were recently identified as vital key regulators of cell mitosis and have consequently been investigated as therapeutic targets in preclinical and clinical studies. Aurora kinase inhibitors (AKI) have been
Johan D Oslob et al.
Bioorganic & medicinal chemistry letters, 18(17), 4880-4884 (2008-08-06)
This communication describes the discovery of a novel series of Aurora kinase inhibitors. Key SAR and critical binding elements are discussed. Some of the more advanced analogues potently inhibit cellular proliferation and induce phenotypes consistent with Aurora kinase inhibition. In

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service