Skip to Content
Merck
All Photos(1)

Key Documents

12-341

Sigma-Aldrich

Rabbit Anti-Chicken IgY Antibody, HRP conjugate

Upstate®, from rabbit

Synonym(s):

Chicken IgG

Sign Into View Organizational & Contract Pricing

Select a Size

500 μG
€278.00

€278.00


Estimated to ship on25 May 2025


Request a Bulk Order

Select a Size

Change View
500 μG
€278.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€278.00


Estimated to ship on25 May 2025


Request a Bulk Order

biological source

rabbit

Quality Level

conjugate

peroxidase conjugate

antibody form

affinity purified immunoglobulin

antibody product type

secondary antibodies

clone

polyclonal

species reactivity

chicken

manufacturer/tradename

Upstate®

technique(s)

ELISA: suitable
immunohistochemistry: suitable
western blot: suitable

shipped in

dry ice

target post-translational modification

unmodified

Related Categories

General description

Chickens transfer high quantities of IgY (IgG) into the egg yolk and harvesting antibodies from eggs eliminates the need for the invasive bleeding procedure. One week′s eggs can contain 10 times more antibodies than the volume of rabbit blood obtained from one weekly bleeding.

However, there are some disadvantages when using certain chicken derived antibodies in immunoassays. Chicken IgY does not fix mammalian complement component C1 and it does not perform as a precipitating antibody using standard solutions.

Application

Detect Chicken IgY using this Rabbit anti-Chicken IgY Antibody, HRP conjugate validated for use in ELISA, WB, IH.
Research Category
Secondary & Control Antibodies
Research Sub Category
Whole Immunoglobulin Secondary Antibodies

Quality

Routinely evaluated by western blot.

Physical form

0.02M Potassium Phosphate, 0.15M NaCl, pH 7.2, 10mg/ml BSA, and 0.01% gentamicin sulfate

Storage and Stability

2 years at 4°C

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pelagia Koukouraki et al.
Open biology, 6(4), 160022-160022 (2016-06-02)
Genetic and biochemical studies have established a central role for α-synuclein (SNCA) accumulation in the pathogenesis of Parkinson's disease. Uncovering and subsequently interfering with physiological mechanisms that control SNCA expression is one approach to limit disease progression. To this end
Abhilash I Chiramel et al.
Cell reports, 27(11), 3269-3283 (2019-06-13)
Tripartite motif-containing protein 5α (TRIM5α) is a cellular antiviral restriction factor that prevents early events in retrovirus replication. The activity of TRIM5α is thought to be limited to retroviruses as a result of highly specific interactions with capsid lattices. In contrast
Zhiyong Zhang et al.
Plant biotechnology journal, 18(4), 1056-1065 (2019-10-05)
Gluten-free foods cannot substitute for products made from wheat flour. When wheat products are digested, the remaining peptides can trigger an autoimmune disease in 1% of the North American and European population, called coeliac disease. Because wheat proteins are encoded
Naomi Matsumoto et al.
Scientific reports, 8(1), 6701-6701 (2018-05-02)
Secretory lysosomes are required for the specialised functions of various types of differentiated cells. In osteoclasts, the lysosomal proton pump V-ATPase (vacuolar-type ATPase) is targeted to the plasma membrane via secretory lysosomes and subsequently acidifies the extracellular compartment, providing optimal
Eva On-Chai Lau et al.
eLife, 10 (2021-04-27)
Diaphanous (DIAPH) three (DIAPH3) is a member of the formin proteins that have the capacity to nucleate and elongate actin filaments and, therefore, to remodel the cytoskeleton. DIAPH3 is essential for cytokinesis as its dysfunction impairs the contractile ring and

Questions

  1. How can we reconstitute this antibody ? 

    1 answer
    1. Reconstitute this product by adding 500µl sterile, distilled water containing 50% glycerol to make a 1mg/ml stock solution. The reconstitution information can be found in the Certificate of Analysis.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service