Skip to Content
Merck
All Photos(1)

Key Documents

119125

Millipore

Adenosine 5′-Triphosphate, Disodium, Trihydrate, Low Metals Grade

Low metals grade. Specially processed to reduce levels of contaminating metals. Suitable for enzyme kinetic studies.

Synonym(s):

Adenosine 5′-Triphosphate, Disodium, Trihydrate, Low Metals Grade, ATP, 2Na

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
Inquire


Estimated to ship on24 May 2025


Request a Bulk Order

Select a Size

Change View
1 G
Inquire

About This Item

Empirical Formula (Hill Notation):
C10H14N5O13P3 · 2Na · 3H2O
CAS Number:
Molecular Weight:
605.19
MDL number:
UNSPSC Code:
12352208
NACRES:
NA.51


Estimated to ship on24 May 2025


Request a Bulk Order

Quality Level

description

Merck USA index - 14, 155

Assay

≥95% (HPLC)

form

crystalline solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)

impurities

≤0.1% inorganic phosphorus

color

white

solubility

water: 10 mg/mL

cation traces

Al3+: ≤20 ppm
Ca: ≤5 ppm
Fe2+: ≤10 ppm
Mg2+: ≤5 ppm
Zn2+: ≤5 ppm

shipped in

ambient

storage temp.

−20°C

SMILES string

[Na+].[Na+].[P](=O)([O-])([O-])O[P](=O)(O[P](=O)(OC[C@H]1O[C@H]([C@@H]([C@@H]1O)O)[n]2c3ncnc(c3nc2)N)O)O

InChI

1S/C10H16N5O13P3.2Na/c11-8-5-9(13-2-12-8)15(3-14-5)10-7(17)6(16)4(26-10)1-25-30(21,22)28-31(23,24)27-29(18,19)20;;/h2-4,6-7,10,16-17H,1H2,(H,21,22)(H,23,24)(H2,11,12,13)(H2,18,19,20);;/q;2*+1/p-2/t4-,6-,7-,10-;;/m1../s1

InChI key

TTWYZDPBDWHJOR-IDIVVRGQSA-L

Related Categories

General description

Specially processed to reduce levels of contaminating metals. Suitable for enzyme kinetic studies.

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Solheim, L.P., and Fromm, H.J. 1980. Anal. Biochem.109, 266.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service