Skip to Content
Merck
All Photos(5)

Key Documents

G7250

Sigma-Aldrich

D-Glucose 6-phosphate disodium salt hydrate

≥98% (HPLC)

Synonym(s):

G-6-P-Na2, Robison ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H11Na2O9P · xH2O
CAS Number:
Molecular Weight:
304.10 (anhydrous basis)
Beilstein:
5199009
EC Number:
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.25

biological source

synthetic (organic)

Quality Level

Assay

≥98% (HPLC)

form

powder

technique(s)

HPLC: suitable

color

white

solubility

water: 50 mg/mL, clear, colorless

cation traces

Na: 13.5-16.7% (anhydrous)

storage temp.

−20°C

SMILES string

O.[Na+].[Na+].OC1O[C@H](COP([O-])([O-])=O)[C@@H](O)[C@H](O)[C@H]1O

InChI

1S/C6H13O9P.2Na.H2O/c7-3-2(1-14-16(11,12)13)15-6(10)5(9)4(3)8;;;/h2-10H,1H2,(H2,11,12,13);;;1H2/q;2*+1;/p-2/t2-,3-,4+,5-,6?;;;/m1.../s1

InChI key

UUWJZXLTPORJKW-WYFATIGWSA-L

Looking for similar products? Visit Product Comparison Guide

Application

D-Glucose 6-phosphate disodium salt hydrate has been used as a component in the reaction mixture to assay reduced nicotinamide adenine dinucleotide phosphate (NADPH) diaphorase activity, NAD(P)H-cytochrome c reductase activity, and ferredoxin (Fd)-dependent cytochrome c reduction activity.

Biochem/physiol Actions

D-Glucose 6-phosphate is the core carbohydrate substrate for the enzymatic synthesis of archaetidyl-myo-inositols produced through a pathway involving myo-inositol 1-phosphate synthase and CDP-archaeol.

Quality

usually 2-4 H2O/mole

Other Notes

To gain a comprehensive understanding of our extensive range of Monosaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hiroyuki Morii et al.
The Journal of biological chemistry, 284(45), 30766-30774 (2009-09-11)
Ether-type inositol phospholipids are ubiquitously distributed in Archaea membranes. The present paper describes a novel biosynthetic pathway of the archaeal inositol phospholipid. To study the biosynthesis of archaetidylinositol in vitro, we prepared two possible substrates: CDP-archaeol, which was chemically synthesized
Daisuke Seo et al.
Biochimica et biophysica acta, 1794(4), 594-601 (2009-01-24)
Ferredoxin-NADP(+) oxidoreductases (FNRs) of Bacillus subtilis (YumC) and Rhodopseudomonas palustris CGA009 (RPA3954) belong to a novel homo-dimeric type of FNR with high amino acid sequence homology to NADPH-thioredoxin reductases. These FNRs were purified from expression constructs in Escherichia coli cells
Khosbayar Lkhagvadorj et al.
International journal of molecular sciences, 22(1) (2020-12-31)
Prenatal smoke exposure (PreSE) is a risk factor for nicotine dependence, which is further enhanced by postnatal smoke exposure (PostSE). One susceptibility gene to nicotine dependence is Cytochrome P450 (CYP) 2A6, an enzyme responsible for the conversion of nicotine to
Federica Pisaneschi et al.
Molecules (Basel, Switzerland), 24(13) (2019-07-22)
We recently reported that SF2312 ((1,5-dihydroxy-2-oxopyrrolidin-3-yl)phosphonic acid), a phosphonate antibiotic with a previously unknown mode of action, is a potent inhibitor of the glycolytic enzyme, Enolase. SF2312 can only be synthesized as a racemic-diastereomeric mixture. However, co-crystal structures with Enolase
Khosbayar Lkhagvadorj et al.
Epigenetics, 15(12), 1370-1385 (2020-06-24)
Prenatal smoke exposure (PSE) is a risk factor for nicotine dependence. One susceptibility gene for nicotine dependence is Cytochrome P450 (CYP) 2A6, an enzyme responsible for the conversion of nicotine to cotinine and nicotine clearance in the liver. Higher activity

Articles

Review the 10 steps of glycolysis in the Embden-Meyerhof-Parnas glycolytic pathway. Easily compare reaction stages and buy the enzymes for your life science research.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service