Skip to Content
Merck
All Photos(1)

Key Documents

118410

Millipore

Adenosine 3ʹ-Phosphate 5ʹ-Phosphosulfate, Tetralithium Salt

Synonym(s):

Adenosine 3ʹ-Phosphate 5ʹ-Phosphosulfate, Tetralithium Salt, PAPS, 4Li

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
€613.00

€613.00


Estimated to ship on23 May 2025


Request a Bulk Order

Select a Size

Change View
10 MG
€613.00

About This Item

Empirical Formula (Hill Notation):
C10H11N5O13P2S · 4Li
CAS Number:
Molecular Weight:
531.00
MDL number:
UNSPSC Code:
12352201
NACRES:
NA.51

€613.00


Estimated to ship on23 May 2025


Request a Bulk Order

Assay

≥80% (HPLC)

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)

color

white

solubility

water: 5 mg/mL

shipped in

wet ice

storage temp.

20-25°C

General description

Note: Unstable at room temperature.
Prepared enzymatically. A substrate for sulfotransferases, including Sulfotransferase, Nod Factor (Cat. No. 574300).
Prepared enzymatically. A sulfate donor substrate for sulfotransferases, including Sulfotransferase, Nod Factor (Cat. No. 574300).

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-70°C). Stock solutions are stable for up to 3 months at -70°C. Note: Unstable at room temperature.

Other Notes

Lin, C.-H., et al. 1995. J. Am. Chem. Soc. 117, 8031.
Mandon, E.C., et al. 1994. Proc. Natl. Acad. Sci. USA 91, 10707.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service