Skip to Content
Merck

Skip To

94455

Adenosine 3′-phosphate 5′-phosphosulfate triethylammonium salt

≥90.0% (HPLC)

Synonym(s):

3′-Phosphoadenosine-5′-phosphosulfate triethylammonium salt

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
1 mg
Check Cart for Availability
€ 221,00
5 mg
Check Cart for Availability
€ 878,00
25 mg
Check Cart for Availability
€ 3.460,00

About This Item

Empirical Formula (Hill Notation):
C10H15N5O13P2S · xC6H15N
CAS Number:
Molecular Weight:
507.26 (free acid basis)
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.32

€ 221,00


Check Cart for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥90.0% (HPLC)

form

powder or crystals

solubility

water: soluble

storage temp.

−20°C

SMILES string

CCN(CC)CC.Nc1ncnc2n(cnc12)[C@@H]3O[C@H](COP(O)(=O)OS(O)(=O)=O)[C@@H](OP(O)(O)=O)[C@H]3O

InChI

1S/C10H15N5O13P2S.C6H15N/c11-8-5-9(13-2-12-8)15(3-14-5)10-6(16)7(27-29(17,18)19)4(26-10)1-25-30(20,21)28-31(22,23)24;1-4-7(5-2)6-3/h2-4,6-7,10,16H,1H2,(H,20,21)(H2,11,12,13)(H2,17,18,19)(H,22,23,24);4-6H2,1-3H3/t4-,6-,7-,10-;/m1./s1

InChI key

IIAWHKVLRMAJSN-MCDZGGTQSA-N

General description

Adenosine 3′-phosphate 5′-phosphosulfate or 3′-Phosphoadenosine-5′-phosphosulfate (PAPS) is produced by the action of PAPS synthase in the cytoplasm and nucleus in animal cells. It is also found in the cytoplasm and plastids of plant cells.

Application

Adenosine 3′-phosphate 5′-phosphosulfate triethylammonium salt has been used in the determination of human 3-O-sulfotransferase-1 (3-OST-1) and aryl-sulfotransferase (AST-IV) enzyme activity.

Biochem/physiol Actions

3′-Phosphoadenosine-5′-phosphosulfate (PAPS) acts as a sulfate donor and serves as a substrate for sulfotransferases (STs).

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
A5763A166A165
assay

≥90.0% (HPLC)

assay

≥96%

assay

≥98% (HPLC)

assay

≥98% (HPLC)

solubility

water: soluble

solubility

water: 25 mg/mL, clear, colorless to very faintly yellow

solubility

H2O: soluble

solubility

H2O: 10 mg/mL

Quality Level

100

Quality Level

200

Quality Level

100

Quality Level

100

form

powder or crystals

form

powder

form

solid

form

powder

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C


pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters