Key Documents
1.17348
Millipore
MAS-100 Atmos® Gas Exhaust Adapter
for use with MAS-100 Atmos®
Select a Size
1173480001
€935.00
€935.00
Estimated to ship on28 May 2025
Request more information
Select a Size
Change View
1173480001
€935.00
About This Item
UNSPSC Code:
41103403
NACRES:
NA.74
€935.00
Estimated to ship on28 May 2025
Request more information
material
stainless steel adapter (
)
feature
autoclavable
manufacturer/tradename
MAS-100 Atmos®
application(s)
compressed gas monitoring
cosmetics
food and beverages
pharmaceutical
compatibility
for use with MAS-100 Atmos®
General description
The MAS-100 Atmos® Gas Exhaust Tube Adapter is an accessory of MAS-100 Atmos® Microbial Compressed Gas Sampler for safe removal of sampled gases to prevent intoxication and contamination of the environment.
The adapter has a customized gas exhaust tubing with a minimum inner diameter of 32 mm and a maximum length of 5 m. These are the conditions validated by the manufacturer.
The connector piece of the Gas Exhaust Tube is connected to the exhaust of the MAS-100 Atmos® instrument. The alternative tubing is fixed to the connector with the tri-clamp.
Application
MAS-100 Atmos® Microbial Compressed Gas Sampler is used to sample pre-defined compressed gases like argon, carbon dioxide, compressed air, and nitrogen from pharmaceutical, cosmetic, food & beverage industry.
Features and Benefits
Removal of potentially toxic gas out of the sampling environment
Legal Information
MAS-100 Atmos is a registered trademark of MBV AG