Key Documents
1.17367
Millipore
MAS-100 Atmos® Tri-Clamp (34 mm)
for use with MAS-100 Atmos®, autoclavable
Select a Size
1173670001
€200.00
€200.00
Estimated to ship on01 June 2025
Request more information
Select a Size
Change View
1173670001
€200.00
About This Item
UNSPSC Code:
41103403
NACRES:
NA.74
€200.00
Estimated to ship on01 June 2025
Request more information
material
stainless steel clamp(s)
feature
autoclavable
manufacturer/tradename
MAS-100 Atmos®
application(s)
compressed gas monitoring
cosmetics
food and beverages
pharmaceutical
compatibility
for use with MAS-100 Atmos®
General description
MAS-100 Atmos® Tri clamp (34 mm) is an accessory designed to tighten the pressure tube connection. It is made from stainless steel.
Place the silicone gasket (ref. 1.17364) on top of the closed sampling head and fix the pressure connection tubing with the 34 mm tri-clamp
Application
MAS-100 Atmos® Tri clamp (34 mm) is used to fix the pressure tube connection with the MAS-100 Atmos® microbial compressed gas sampler.
Legal Information
MAS-100 Atmos is a registered trademark of MBV AG