Skip to Content
Merck
All Photos(1)

Key Documents

1.17084

Millipore

MAS-100 Iso NT® Silicone Gasket

with standard base, for use with MAS-100® Iso, pkg of 5 pieces

Sign Into View Organizational & Contract Pricing

Select a Size

1 PKG
€157.00

€157.00


Estimated to ship on10 May 2025


Request more information

Select a Size

Change View
1 PKG
€157.00

About This Item

UNSPSC Code:
41122409
NACRES:
NA.74

€157.00


Estimated to ship on10 May 2025


Request more information

material

silicone gasket

Agency

non-compendial

feature

disposable

packaging

pkg of 5 pieces

application(s)

air monitoring
pharmaceutical

compatibility

for use with MAS-100® Iso

shipped in

ambient

storage temp.

no temp limit

General description

MAS-100 ISO NT® Silicone Gasket is an additional accessory designed to be specifically used for MAS-100 ISO NT® and MAS-100 ISO MH®. The MAS 100 Iso family microbial air samplers have been specially developed for monitoring and measuring the microbiological contamination at fixed sampling locations in cleanrooms, RABS and isolators. The Silicone Gasket is made of transparent Platinum cured silicone and can be sterilized by autoclaving. The silicone gasket is used to seal the tri clamp connections on instruments of the MAS-100 Iso product family. The surface of the gasket is smooth and nonporous, free of laminar recesses and increases, edges without tears and notches.

Application

MAS-100 ISO NT® Silicone Gasket is an additional accessory used to seal the tri clamp connections on to the instrument. MAS-100 Iso family devices are extensively used in Pharmaceutical applications.

Legal Information

MAS-100 is a registered trademark of MBV AG
MAS-100 ISO MH is a registered trademark of MBV AG
MAS-100 ISO NT is a registered trademark of MBV AG

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service