Skip to Content
Merck

1.16887

Millipore

Fractogel® EMD TMAE (S)

Synonym(s):

Biochromatography, Fractogel® Ion exchangers, Ion exchanger for LC (Fractogel® TSK/EMD), Fractogel® EMD TMAE (S)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115711
Pricing and availability is not currently available.

ligand

(Trimethylammoniumethyl)

Quality Level

description

strong anion exchanger, suspension in 20% ethanol and 150 mM NaCl (20-40 µm)

sterility

sterile (Caustic Stable)

product line

Fractogel®

form

resin

parameter

8 bar max. pressure
80 cm/hr flow rate

matrix active group

methacrylate

capacity

100 mg binding capacity (BSA/mg of resin)

transition temp

flash point 35 °C (calculated)

density

1.430 g/cm3 at 20 °C

bulk density

1000 kg/m3

application(s)

gene therapy
recombinant protein
vaccine development

separation technique

anion exchange
strong anion exchange

storage temp.

2-30°C

Looking for similar products? Visit Product Comparison Guide

General description

Stron anion exchange Fractogel® resin with small particle size, featuring the tentacle technology with a trimethylammoniumethyl functional group, for purification of acidic and neutral proteins and peptides from multiple sources, DNA and virus removal, plasmid isolation, vaccine purification, and more

Features and Benefits

Fractogel® EMD TMAE (S) (20-40 µm particle size) enables:

High protein loading conditions resulting in good selectivity and resolution with the tentacle technology
  • Excellent binding to large viruses at increased loading
  • Homogenous binding with high selectivity and purity
  • Reduce operating costs and resin replacement with enduring durability - allowing for multiple cycles of column regeneration and sanitization
  • Compatibility with 2.5 % (v/v) aqueous benzyl alcohol containing 150 mM NaCl storage solution

Packaging

  • 1.16887.0100: Fractogel® EMD TMAE (S) Resin 100ml
  • 1.16887.0010: Fractogel® EMD TMAE (S) Resin 10ml
  • 1.16887.0500: Fractogel® EMD TMAE (S) Resin 500ml
  • 1.16887.5000: Fractogel® EMD TMAE (S) Resin 5L

Plastic bottle

Analysis Note

Appearance: Milky turbid suspension, free from impurities (foreign particles)
Microscopic evaluation: Uniform spherical particles, no agglomerates, no fines
Extractable matter (water): ≤ 0.03 %
Ce content: ≤ 1 µg/g
Pressure drop(column: ID=1.6 cm, L=10 cm at 5 ml/min): ≤ 5.0 bar
Particle size (d10): 20 - 28 µm
Particle size (d50): 24 - 34 µm
Particle size (d90): 28 - 38 µm
Colony forming units (TAMC + TYMC): ≤ 100 CFU/ml
Endotoxins: ≤ 1.00 EU/ml
Protein binding capacity (bovine serum albumin): 80 - 120 mg/ml
Functional test (b:a): ≤ 0.1
Functional Test: Seperation of conalbumin and human serum albumin

Legal Information

FRACTOGEL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

95.0 °F

Flash Point(C)

35 °C


What is the Emprove® Program?

The Emprove® Program is a system providing comprehensive and thorough documentation of our filters and single-use components, pharma raw materials, and starting materials. Four document types are included with an Emprove® Program subscription:

Material Qualification Dossier

Free of charge - Sign in to download

Quality Management Dossier (Open Access)

Free of charge - Sign in to download

Operational Excellence Dossier

Available by subscription or for a fee

Learn more about the benefits an Emprove® Program subscription can provide
Why are these documents unavailable?
Emprove® Dossiers contain information intended to support the registration process and risk assessment activities of pharmaceutical producers and contain confidential information. Please contact us if you would like access.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Explore strategies for separating empty and filled capsids in viral vector production. Learn about upstream strategies and how trends in AAV capsid design impact purification.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service