Skip to Content
AT
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Fractogel
®
EMD TMAE (S)
Products
Product Category
columns (4)
Brand
Millipore (4)
Physical Form
Product Line
Separation Technique
ID (mm)
Volume (ml)
Application
Housing Material
Particle Size (µm)
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
1.25071
Fractogel
®
EMD TMAE (S),
MiniChrom Column Fractogel
®
TMAE (S), volume 5 mL, prepacked column
Expand
View Pricing
125057
Fractogel
®
EMD TMAE (S),
MiniChrom Column Fractogel
®
TMAE (S), volume 1 mL, prepacked column
Expand
View Pricing
1.25086
Fractogel
®
EMD TMAE (S),
RoboColumn Fractogel
®
TMAE (S), volume 0.2 mL, prepacked column
Expand
View Pricing
1.25138
Fractogel
®
EMD TMAE (S),
RoboColumn Fractogel
®
TMAE (S), volume 0.6 mL, prepacked column
Expand
View Pricing