1.15419
Cetyl palmitate 15 (cetylesters wax)
EMPROVE® ESSENTIAL, Ph. Eur., NF
Pharma Manufacturing
Synonym(s):
Cetyl palmitate 15 (cetylesters wax), Cetaceum, Spermaceti synthetic
About This Item
Agency
NF
Ph. Eur.
Quality Level
product line
EMPROVE® ESSENTIAL
form
solid
mp
43-57 °C
application(s)
pharmaceutical
semi-solid formulation
storage temp.
15-25°C
SMILES string
O(CCCCCCCCCCCCCCCC)C(=O)CCCCCCCCCCCCCCC
InChI
1S/C32H64O2/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-29-31-34-32(33)30-28-26-24-22-20-18-16-14-12-10-8-6-4-2/h3-31H2,1-2H3
InChI key
PXDJXZJSCPSGGI-UHFFFAOYSA-N
General description
As part of our Emprove® Program, our raw materials are offered with extensive documentation facilitating compliance of your pharma and biopharma product, full supply chain transparency and risk mitigation. Our SAFC® portfolio of high-quality products for biopharmaceutical and pharmaceutical formulation and production withstands strict quality control procedures and is produced according to applicable cGMP guidelines.
Application
Legal Information
Application
related product
Storage Class Code
11 - Combustible Solids
WGK
nwg
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service