Skip to Content
Merck
All Photos(1)

Key Documents

1.15096

Supelco

Extrelut® NT 20 Pre-packed SLE Column

20 ml sample size

Synonym(s):

TM=["EXtrelut"] NT 20

Sign Into View Organizational & Contract Pricing

Select a Size

25 UNITS
€542.00

€542.00


Estimated to ship on28 May 2025


Request a Bulk Order

Select a Size

Change View
25 UNITS
€542.00

About This Item

UNSPSC Code:
41115711
NACRES:
NC.07

€542.00


Estimated to ship on28 May 2025


Request a Bulk Order

Product Name

EXtrelut® NT 20, pre-packed columns for extraction of lipophilic compounds from aqueous solutions (20 ml sample size)

Quality Level

technique(s)

supported liquid extraction (SLE): suitable

storage temp.

15-25°C

General description

Extrelut® NT simplifies liquid-liquid extraction in its most effective form by using a chemically inert, wide-pore and highly pure diatomaceous earth based solid phase. Depending on the sample volume three types of columns are available: Extrelut NT1 can take up a maximum of one ml, Extrelut NT3 three ml and Extrelut NT20 up to twenty ml of sample.
pre-packed columns for extraction of lipophilic compounds from aqueous solutions (20 ml sample)

Application


  • Simultaneous determination of ethyl carbamate and chloropropanols in flavorings by gas chromatography-triple quadrupole tandem mass spectrometry: This study utilized Extrelut NT 20 for solid-phase extraction in the analysis of volatile compounds, highlighting its effectiveness in complex sample preparations (Xu et al., 2013).

  • Determination of 10 heterocyclic aromatic amines in beef jerky by high performance liquid chromatography: Extrelut NT 20 was used for extracting aromatic amines from processed food samples, demonstrating its utility in food safety applications (Wan et al., 2012).

  • Determination of chloropropanols in foods by one-step extraction and derivatization using pressurized liquid extraction and gas chromatography-mass spectrometry: This research showcases the adaptability of Extrelut NT 20 in streamlining the preparation steps for environmental contaminant analysis (Racamonde et al., 2011).

  • Confirmatory analysis of acetylgestagens in plasma using liquid chromatography-tandem mass spectrometry: The use of Extrelut NT 20 in this study underlines its critical role in the precise and reliable measurement of pharmaceutical compounds in biological matrices (Mortensen and Pedersen, 2007).

  • Simultaneous quantitative analysis of ketanserin and ketanserinol in plasma by RP-HPLC with fluorescence detection: Demonstrates the application of Extrelut NT 20 in therapeutic drug monitoring, ensuring accurate pharmacokinetic assessments (Yassen et al., 2003).

Analysis Note

filling volume: difference max. 5% from ideal filling volume
purity test
- absorbance 280 nm:
≤ 0.040
- absorbance 250 nm: ≤ 0.080
- absorbance 230 nm: ≤ 0.130

Legal Information

Extrelut is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

STOT RE 1 Inhalation

Target Organs

Lungs

Storage Class Code

6.1D - Non-combustible, acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Simplify your liquid-liquid extractions with our emulsion-free supported-liquid extraction (SLE) single step solution.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service