Skip to Content
Merck
All Photos(1)

Key Documents

1.15029

Millipore

M 17 broth acc. to TERZAGHI

for microbiology

Sign Into View Organizational & Contract Pricing

Select a Size

500 G
€352.00

€352.00


Estimated to ship on22 April 2025


Request more information

Select a Size

Change View
500 G
€352.00

About This Item

UNSPSC Code:
41106213
NACRES:
NA.74

€352.00


Estimated to ship on22 April 2025


Request more information

Quality Level

form

medium granules (dehydrated (DCM))

manufacturer/tradename

GranuCult®

technique(s)

microbiological culture: suitable

pH

7.1-7.3 (25 °C, 42.5 g/L in H2O, after autoclaving)

solubility

42.5 g/L

bulk density

530 kg/m3

application(s)

food and beverages
microbiology

storage temp.

15-25°C

suitability

Lactobacillus spp.

Analysis Note

Appearance (clearness): clear
Appearance (colour): brown
pH-value (25 °C): 7.0 - 7.4
Growth (Streptococcus agalactiae ATCC 13813): good to very good
Growth (Lactococcus lactis spp. cremoris ATCC 19257): good to very good
Growth (Lactococcus lactis spp. lactis ATCC 19435 (WDCM 00016)): good to very good
Growth (Enterococcus faecalis ATCC 11700): good to very good
Growth (Escherichia coli ATCC 25922 (WDCM 00013)): good to very good
Growth (Staphylococcus aureus ATCC 25923 (WDCM 00034)): good to very good
Growth (Lactobacillus acidophilus ATCC 4356 (WDCM 00098)): good to very good
Growth (Lactobacillus casei ATCC 393 (WDCM 00100)): good to very good
Growth (Lactobacillus fermentum ATCC 9338): fair to very good
Incubation: 48 hrs.; 28 °C; aerobic

Footnote

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service