Skip to Content
Merck
All Photos(1)

Key Documents

1.14942

Supelco

Nitrate Test, photometric

photometric, 0.2-17.0 mg/L (NO3-N), 0.9-75.3 mg/L (NO3-), Spectroquant®

Sign Into View Organizational & Contract Pricing

Select a Size

50 TESTS
€161.00

€161.00


Estimated to ship on23 May 2025


Request a Bulk OrderRequest more information

Select a Size

Change View
50 TESTS
€161.00

About This Item

UNSPSC Code:
41116116
NACRES:
NA.24

€161.00


Estimated to ship on23 May 2025


Request a Bulk OrderRequest more information

Product Name

Nitrate test in seawater, photometric, 0.2-17.0 mg/L (NO3-N), 0.9-75.3 mg/L (NO3-), Spectroquant®

product line

Spectroquant®

Quality Level

usage

sufficient for 50 tests

specific analyte(s)

nitrate

measuring range

0.2-17.0 mg/L (NO3-N)
0.9-75.3 mg/L (NO3-)

technique(s)

photometry: suitable

compatibility

for use with Spectroquant® Nova 60 A
for use with Spectroquant® Prove 100
for use with Spectroquant® Prove 300
for use with Spectroquant® Prove 600

detection method

photometric (Resorcinol)

storage temp.

15-25°C

General description

Method: photometric 0.2 - 17.0 mg/l NO₃-N 0.9 - 75.3 mg/l NO₃⁻ Spectroquant®
This Spectroquant® Nitrate Cell Test allows the accurate quantification of nitrate in seawater samples. Nitrate is an essential nutrient required for the growth of animals and plants. However, elevated levels of nitrate can be a health risk, in particular for pregnant women and infants. Therefore, nitrate levels in water sources are regulated in many countries.
Method applied: In strongly sulfuric solution, in the presence of chloride nitrate ions react with resorcinol to form a red-violet indophenol dye that is determined photometrically.
The Spectroquant® Reagent Test Kits contain highly stable, ready-to-use reagent mixtures to perform the analysis according to the procedure described in the accompanying instruction leaflet.
All our Cell and Reagent Test Kits are equipped with the unique Live ID (2D barcode) which allows seamless method recognition and contains essential information such as lot number, expiry date, and automatic calibration updates.

Legal Information

SPECTROQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 3 - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1A - Skin Sens. 1 - STOT SE 1 Oral - STOT SE 2 Oral

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Hoang Nhat Phong Vo et al.
Bioresource technology, 303, 122877-122877 (2020-02-07)
This study investigated the extracellular polymeric substance (EPS) and enzyme extrusion of Chlorella sp. using seven carbon sources and two salinities for potential pollutant co-metabolism. Results indicated that the levels of biomass, EPS and enzymes of microalgae cultured with glucose

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service