Skip to Content
Merck

Skip To

1.14657

Ammonium Test Kit, colorimetric

0.5-10 mg/L (NH₄⁺), MQuant®

Synonym(s):

Ammonium Visual Test Kit

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
50 tests
Check Cart for Availability
€ 70,50

About This Item

UNSPSC Code:
41116116
NACRES:
NA.24

€ 70,50


Check Cart for Availability

Request a Bulk OrderRequest more information
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Ammonium Test in freshwater and seawater, colorimetric, 0.5-10 mg/L (NH4+), for use with MQUANT®

Quality Segment

specific analyte(s)

ammonium

measuring range

0.5-10 mg/L (NH4+)

compatibility

for use with MQUANT®

detection method

colorimetric

storage temp.

15-25°C

General description

Method: colorimetric with color card 0.5 - 1 - 2 - 3 - 5 - 10 mg/l NH₄⁺ MQuant®
The test measures both ammonium ions and dissolved ammonia in freshwater and seawater. The test kit contains the easy to use colour card.

Legal Information

MQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
1.080241.144281.14423
specific analyte(s)

ammonium

specific analyte(s)

ammonium

specific analyte(s)

ammonium

specific analyte(s)

ammonium

detection method

colorimetric

detection method

colorimetric

detection method

colorimetric

detection method

colorimetric

measuring range

0.5-10 mg/L (NH4+)

measuring range

0.2-5 mg/L (NH4+)

measuring range

0.025-0.4 mg/L (NH4+)

measuring range

0.16-6.2 mg/L (NH4-N), 0.2-8 mg/L (NH4+)

compatibility

for use with MQUANT®

compatibility

for use with MQUANT®

compatibility

for use with MQuant®

compatibility

for use with MQUANT®

storage temp.

15-25°C

storage temp.

15-25°C

storage temp.

15-25°C

storage temp.

15-25°C

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100


Still not finding the right product?


signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Flam. Liq. 2 - Met. Corr. 1 - Skin Corr. 1A - STOT SE 3

target_organs

Central nervous system

Storage Class

3 - Flammable liquids

wgk

WGK 3



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters