Skip to Content
Merck
All Photos(2)

Key Documents

1.14423

Supelco

Ammonium Test Kit, colorimetric

0.2-8 mg/L (NH₄⁺), 0.16-6.2 mg/L (NH₄-N), MQuant®

Synonym(s):

Ammonium Visual Test Kit

Sign Into View Organizational & Contract Pricing

Select a Size

200 TESTS
€136.00

€136.00


Estimated to ship on29 May 2025


Request a Bulk OrderRequest more information

Select a Size

Change View
200 TESTS
€136.00

About This Item

UNSPSC Code:
41116116
NACRES:
NA.24

€136.00


Estimated to ship on29 May 2025


Request a Bulk OrderRequest more information

Product Name

Ammonium Test, colorimetric, 0.2-8 mg/L (NH4+), 0.16-6.2 mg/L (NH4-N), for use with MQUANT®

specific analyte(s)

ammonium

Quality Level

measuring range

0.16-6.2 mg/L (NH4-N)
0.2-8 mg/L (NH4+)

compatibility

for use with MQUANT®

detection method

colorimetric

storage temp.

15-25°C

General description

Method: colorimetric, indophenol blue, with color card and comparator block with short test tubes 0.2-0.5-0.8-1.2-1.6-2-3-5-8 mg/l NH₄⁺ 0.16-0.4-0.6-0.9-1.2-1.6-2.3-3.9-6.2 mg/l NH₄-N MQuant®
With the MQuant® Ammonium test (formally MColortest®) ammonium ions can be detected in e.g. groundwater and surface water, seawater, drinking water and aquarium water. Ammonium nitrogen (NH4-N) occurs partly in the form of ammonium ions and partly as ammonia. A pH-dependent equilibrium exists between the two forms.

Method
In strongly alkaline solution ammonia reacts with a chlorinating agent to form monochloramine. This in turn reacts with thymol to form a blue indophenol derivative. Due to the intrinsic yellow coloration of the reagent blank, the measurement solution is yellow-green to green in color. The ammonium concentration is measured semiquantitatively by visual comparison of the color of the measurement solution with the color fields of a color card. The unique brilliance of the print and their fine color graduation allow precise analyses, even at the lower end of the concentration range. Using the refill pack 1.18455 the original test kit can be used for a very long time, making it a particularly economical and ecological system. 

The test is suitable for ammonium determination in
  • Groundwater and surface water
  • Drinking water and mineral water
  • Aquarium water, waters from aquaculture
  • Boiler and boiler feed water, cooling water
  • Process water
  • Waste water
  • Electroplating wastewater
  • Swimming-pool water
  • Nutrient solutions for fertilization
  • Food after appropriate sample pretreatment

This test is not suited for seawater.

Application


  • Evaluation of Water Ammonium Ion Test Kit and Its Feasibility for the Analysis of Protein in Food.: This study evaluates the efficacy of an ammonium ion test kit for water quality analysis and its potential application in determining protein content in food samples, showcasing its versatility in both environmental and food sciences (Wu D et al., 2022).

  • Technical note: Evaluation of a colorimetric point-of-care test for measuring urine ammonium concentration in periparturient dairy cattle.: This research highlights the use of a colorimetric ammonium test for monitoring the health and metabolic status of dairy cattle, demonstrating its practical application in veterinary medicine (Megahed AA, Constable PD, 2020).

  • Using Cellulolytic Nitrogen Fixing Bacterium, Azomonas agilis for Effective Degradation of Agricultural Residues.: Ammonium tests are used to monitor nitrogen fixation and ammonia production in microbial degradation studies, emphasizing their role in sustainable agriculture and waste management (Latt ZK et al., 2018).

  • [Examination of drugs and poisons in institutions not equipped with an analyzer of them and poisons--focusing on the simple qualitative test of surfactants].: This study demonstrates the use of simple ammonium tests for the qualitative analysis of surfactants in forensic and toxicological investigations, highlighting their importance in settings lacking advanced analytical instruments (Kojima Y et al., 2003).

  • [15N] ammonium test for liver function diagnosis.: The [15N] ammonium test is utilized to assess liver function by monitoring nitrogen metabolism, providing valuable insights into liver health and the impact of various treatments on hepatic function (Jung K et al., 1989).

Legal Information

MCOLORTEST is a registered trademark of Merck KGaA, Darmstadt, Germany
MQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Flam. Liq. 2 - Met. Corr. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

71.6 °F

Flash Point(C)

22 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service