Skip to Content
Merck
All Photos(1)

Key Documents

1.14449

Supelco

Phosphate Test Kit, colorimetric

1.0-40 mg/L (PO₄-P), 3.1-123 mg/L (PO₄³⁻), MQuant®

Synonym(s):

Phosphate Visual Test Kit

Sign Into View Organizational & Contract Pricing

Select a Size

190 TESTS
€170.00

€170.00


Estimated to ship on28 May 2025


Request a Bulk OrderRequest more information

Select a Size

Change View
190 TESTS
€170.00

About This Item

UNSPSC Code:
41116116
NACRES:
NA.24

€170.00


Estimated to ship on28 May 2025


Request a Bulk OrderRequest more information

Product Name

Phosphate Test, colorimetric, 1.0-40 mg/L (PO4-P), 3.1-123 mg/L (PO43-), for use with MQUANT®

specific analyte(s)

phosphate

Quality Level

measuring range

1.0-40 mg/L (PO4-P)
3.1-123 mg/L (PO43-)

compatibility

for use with MQUANT®

detection method

colorimetric

storage temp.

15-25°C

General description

Method: colorimetric, VM, with color card and comparator block with short test tubes 1.0 - 2.0 - 3.5 - 6.0 - 10 - 20 - 40 mg/l PO₄-P 3.1 - 6.1 - 11 - 18 - 31 - 61 - 123 mg/l PO₄³⁻ MQuant®

Application


  • Environmental Monitoring: Phosphate tests are extensively used in environmental monitoring to assess the levels of phosphate in water bodies. These tests help in evaluating the impact of agricultural runoff, industrial discharge, and wastewater treatment on water quality. Recent studies have focused on using selective colorimetric nano-sensing solutions to determine phosphate ions in drinking water samples, highlighting their importance in ensuring safe and clean water supplies (Salem and Draz, International Journal of Environmental Analytical Chemistry, 2020).

Legal Information

MQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1A

Storage Class Code

8B - Non-combustible, corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service