Skip to Content
Merck

Skip To

1.10428

Phosphate Test Strips, colorimetric

greener alternative

10-500 mg/L (PO₄³⁻), MQuant®

Synonym(s):

Phosphate pH stick

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
100 tests
Check Cart for Availability
€ 79,20

About This Item

UNSPSC Code:
41116116
NACRES:
NA.24

€ 79,20


Check Cart for Availability

Request a Bulk OrderRequest more information
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Phosphate Test, colorimetric, 10-500 mg/L (PO43-), MQuant®

description

semi-quantitative

Quality Segment

product line

MQuant®

specific analyte(s)

phosphate

greener alternative product characteristics

Waste Prevention
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

measuring range

10-500 mg/L (PO43-)

application(s)

agriculture
food testing
wastewater

compatibility

Reflectoquant®, Spectroquant®

detection method

colorimetric

greener alternative category

storage temp.

15-25°C

General description

Method: Colorimetric, with test strips and reagent, 10 - 25 - 50 - 100 - 250 - 500 mg/L (PO43-), MQuant®

The MQuant® Phosphate test includes test strips and all necessary additional reagents. It is suitable for determining orthophosphate in wastewater, soil samples, fertilizers and food. In sulfuric solution orthophosphate ions (PO43-) react with molybdate ions to form molybdophosphoric acid, which is reduced to phosphomolybdenum blue (PMB). The phosphate concentration is measured semi-quantitatively by visual comparison of the reaction zone of the test strip with the fields of a color scale. The test strips are perfect for pre-testing a sample in an economic and quick way to estimate the phosphate concentration and a possible dilution factor prior to using a quantitative method such as a Reflectoquant® or Spectroquant® Phosphate test.

Design for sustainability benefits:

We continue to demonstrate our commitment to our planet and a sustainable future by exploring ways to improve existing products in our portfolio. We have recently reviewed the current line of MQuant® test strips to identify key opportunities to minimize impact without compromising on the product performance.

This product has benefited from:

  • Test strip with reduction
  • Recycled aluminum tube
  • Digital instruction leaflet
Learn more about the initiative: Small Changes to Test Strips for Big Impact.

Application


  • Zinc and Silicon Nano-Fertilizers Influence Ionomic and Metabolite Profiles in Maize to Overcome Salt Stress: This research utilizes phosphate tests in the study of plant physiology, specifically to gauge the efficacy of nano-fertilizers in enhancing phosphate uptake under salt stress conditions, providing insights into sustainable agricultural practices (Shoukat A et al., 2024).
  • Investigation of Mechanical Properties of Polymer-Infiltrated Tetrapodal Zinc Oxide in Different Variants: Applies phosphate testing in the field of materials science to assess the interaction of phosphate with novel composite materials, thereby informing the development of more durable and efficient materials for various industrial applications (Scherer F et al., 2024).

Legal Information

MQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany
REFLECTOQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany
SPECTROQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
1.148461.169781.18388
detection method

colorimetric

detection method

colorimetric

detection method

reflectometric

detection method

colorimetric

specific analyte(s)

phosphate

specific analyte(s)

phosphate

specific analyte(s)

phosphate

specific analyte(s)

phosphate

compatibility

Reflectoquant®, Spectroquant®

compatibility

for use with MQUANT®

compatibility

RQflex® 20, for use with REFLECTOQUANT®

compatibility

for use with MQUANT®

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

product line

MQuant®

product line

-

product line

-

product line

-

sustainability

Greener Alternative Product

sustainability

-

sustainability

-

sustainability

-


pictograms

Corrosion

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Met. Corr. 1 - Skin Irrit. 2

Storage Class

8A - Combustible, corrosive hazardous materials

wgk

WGK 2



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters