Skip to Content
Merck
All Photos(1)

Key Documents

1.14408

Supelco

Nitrite Test Kit, colorimetric

0.005-0.10 mg/L (NO₂⁻), 0.0015-0.030 mg/L (NO₂-N), MQuant®

Synonym(s):

Nitrite Visual Test Kit

Sign Into View Organizational & Contract Pricing

Select a Size

110 TESTS
€160.00

€160.00


Estimated to ship on17 May 2025


Request a Bulk OrderRequest more information

Select a Size

Change View
110 TESTS
€160.00

About This Item

UNSPSC Code:
41116116
NACRES:
NA.24

€160.00


Estimated to ship on17 May 2025


Request a Bulk OrderRequest more information

Product Name

Nitrite Test, colorimetric, 0.005-0.10 mg/L (NO2-), 0.0015-0.030 mg/L (NO2-N), for use with MQUANT®

specific analyte(s)

nitrite

Quality Level

measuring range

0.0015-0.030 mg/L (NO2-N)
0.005-0.10 mg/L (NO2-)

compatibility

for use with MQUANT®

detection method

colorimetric

storage temp.

15-25°C

General description

Method: colorimetric with color card and comparator block with long test tubes 0.005-0.012-0.02-0.03-0.04-0.05-0.06-0.08-0.10 mg/l NO₂⁻ 0.0015-0.0037-0.006-0.009-0.012-0.015-0.018-0.024-0.030 mg/l NO₂-N MQuant®

Legal Information

MQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service