Skip to Content
Merck
All Photos(1)

Key Documents

1.12516

Sigma-Aldrich

Yttrium(III) nitrate hexahydrate

99+

Synonym(s):

Yttrium(III) nitrate hexahydrate

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
€80.20

€80.20


Estimated to ship on28 March 2025


Request a Bulk Order

Select a Size

Change View
25 G
€80.20

About This Item

Linear Formula:
Y(NO3)3 · 6 H2O
CAS Number:
Molecular Weight:
383.01
MDL number:
UNSPSC Code:
12352302
EC Index Number:
233-802-6
NACRES:
NA.22
form:
solid
solubility:
2304 g/L

€80.20


Estimated to ship on28 March 2025


Request a Bulk Order

Quality Level

form

solid

potency

3890 mg/kg LD50, oral (Rat)

pH

3.5 (20 °C, 50 g/L in H2O)

mp

51.8 °C

solubility

2304 g/L

density

1.1 g/cm3 at 20 °C

bulk density

1100 kg/m3

storage temp.

no temp limit

SMILES string

[Y+3].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.O

InChI

1S/3NO3.6H2O.Y/c3*2-1(3)4;;;;;;;/h;;;6*1H2;/q3*-1;;;;;;;+3

InChI key

QBAZWXKSCUESGU-UHFFFAOYSA-N

Analysis Note

Assay (complexometric; calculated on dried substance): ≥ 99.0 %
Identity (ICP-OES): passes test

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service