Skip to Content
Merck
All Photos(3)

Documents

237957

Sigma-Aldrich

Yttrium(III) nitrate hexahydrate

99.8% trace metals basis

Synonym(s):

Yttrium hexahydrate trinitrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Y(NO3)3 · 6H2O
CAS Number:
Molecular Weight:
383.01
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

99.8% trace metals basis

form

crystals and lumps

impurities

≤2500.0 ppm Trace Rare Earth Analysis

density

2.682 g/mL at 25 °C (lit.)

SMILES string

O.O.O.O.O.O.[Y+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O

InChI

1S/3NO3.6H2O.Y/c3*2-1(3)4;;;;;;;/h;;;6*1H2;/q3*-1;;;;;;;+3

InChI key

QBAZWXKSCUESGU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Yttrium(III) nitrate hexahydrate is a white crystalline solid widely used as a precursor to synthesize yttrium-based complexes and nanomaterials.

Application

Yttrium(III) nitrate hexahydrate can be used:
  • As a precursor to synthesize yttrium oxide nanoparticles by co-precipitation method.
  • To prepare electroactive poly(vinylidene fluoride) (PVDF) thin films. The addition of Yttrium(III) nitrate hexahydrate improves dielectric constant and electroactive β phase nucleation in PVDF films.
  • As a dopant to prepare cerium oxide hollow sphere hierarchical structures with enhanced photocatalytic activity.
  • As a green catalyst for rapid synthesis of supramolecules such as calix[4]resorcinarenes.

Other Notes

Hydration may vary slightly

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rushikesh Fopase et al.
Materials science & engineering. C, Materials for biological applications, 116, 111163-111163 (2020-08-19)
Exclusive magnetocaloric properties of orthoferrites offer advantages for their application in the magnetic hyperthermia as well as imaging applications. In the present study, the effect of yttrium concentration on the magnetic characteristics of the iron oxide based nanomaterials was analyzed
Wang, X., et al.
Nanjing Ligong Daxue Xuebao, 75, 636-636 (2003)
Synthesis, optical and electrochemical properties of Y2O3 nanoparticles prepared by co-precipitation method
Thulasingam Saravanan, et al.
Journal of Nanoscience and Nanotechnology, 15, 4353-4357 (2015)
Yttrium (III) Nitrate: A Powerful Catalyst for Green and Rapid Synthesis of Some Supramolecules
Afsaneh Arami, et al.
Journal of the Chinese Chemical Society, 62, 13-16 (2015)
The role of cerium (iii)/yttrium (iii) nitrate hexahydrate salts on electroactive beta phase nucleation and dielectric properties of poly (vinylidene fluoride) thin films
Pradip Thakur, et al.
Royal Society of Chemistry Advances, 5, 28487-28496 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service