Skip to Content
Merck
All Photos(1)

Key Documents

1.12171

Sigma-Aldrich

Gallium(III) nitrate hydrate 98+

Synonym(s):

Gallium(III) nitrate hydrate 98+

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€144.00

€144.00


Estimated to ship on18 May 2025


Request a Bulk Order

Select a Size

Change View
5 G
€144.00

About This Item

Linear Formula:
Ga(NO3)3 · xH2O
CAS Number:
Molecular Weight:
255.74 (anhydrous basis)
MDL number:
UNSPSC Code:
12352302
EC Index Number:
236-815-5
NACRES:
NA.22
grade:
for analytical purposes
form:
solid

€144.00


Estimated to ship on18 May 2025


Request a Bulk Order

grade

for analytical purposes

Quality Level

form

solid

pH

2 (20 °C, 50 g/L in H2O)

mp

65 °C

storage temp.

no temp limit

SMILES string

[Ga](O[N+](=O)[O-])(O[N+](=O)[O-])O[N+](=O)[O-].O

InChI

1S/Ga.3NO3.H2O/c;3*2-1(3)4;/h;;;;1H2/q+3;3*-1;

InChI key

YVFORYDECCQDAW-UHFFFAOYSA-N

Analysis Note

Assay (complexometric, calc. on the ignited basis): ≥ 98.0 %
Ga (Gallium): 15.0 - 25.0 %
Identity (ICP-OES): passes test

Pictograms

Flame over circleExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Ox. Sol. 2 - Skin Irrit. 2

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service