Skip to Content
Merck
All Photos(2)

Key Documents

254215

Sigma-Aldrich

Indium(III) nitrate hydrate

99.999% trace metals basis

Synonym(s):

Indium trinitrate hydrate, Indium(3+) trinitrate hydrate

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
€116.00
50 G
€460.00

€116.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 G
€116.00
50 G
€460.00

About This Item

Linear Formula:
In(NO3)3 · xH2O
CAS Number:
Molecular Weight:
300.83 (anhydrous basis)
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
form:
crystals and lumps

€116.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

99.999% trace metals basis

form

crystals and lumps

reaction suitability

reagent type: catalyst
core: indium

impurities

≤15.0 ppm Trace Metal Analysis

SMILES string

[In+3].[H]O[H].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O

InChI

1S/In.3NO3.H2O/c;3*2-1(3)4;/h;;;;1H2/q+3;3*-1;

InChI key

YZZFBYAKINKKFM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Indium(III) nitratehydrate is used in the preparation of indium(III) oxide nanoparticles and otherindium complexes.

Application

Indium(III) nitrate hydrate can be used as a precursor to synthesize:
  • In2O3 nanoparticles via thermal decomposition. In2O3 is applicable as a catalyst for reverse water gas shift (RWGS) and CO2 conversion reactions.
  • AgInSe2 thin film on Si substrate via sol-gel spin coating technique.
  • Indium(III)complexes containing thiosemicarbazone. These complexes can be used in the fields of colorimetric and fluorescent probes.

Pictograms

Flame over circleExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Ox. Sol. 2

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Colloidal quantum dots (CQDs) are semiconducting crystals of only a few nanometers (ca. 2–12 nm) coated with ligand/surfactant molecules to help prevent agglomeration.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service