Skip to Content
Merck
All Photos(1)

Key Documents

1.11781

Millipore

Listeria selective enrichment supplement

for Listeria spp., pkg of 10 vials, for use with Listeria-Enrichment-Broth (Base), for use with GranuCult® Buffered Listeria Enrichment Broth (base)

Synonym(s):

Listeria selective supplement

Sign Into View Organizational & Contract Pricing

Select a Size

10 VIAL
€188.00

€188.00


Estimated to ship on28 May 2025


Request more information

Select a Size

Change View
10 VIAL
€188.00

About This Item

UNSPSC Code:
41106212
NACRES:
NA.74

€188.00


Estimated to ship on28 May 2025


Request more information

Agency

(IDF-FIL)

Quality Level

reg. compliance

FDA -BAM

form

solid

composition

Acriflavine HCl, 5 mg per vial
Cycloheximide, 25 mg per vial
Nalidixic acid (sodium salt), 20 mg per vial

packaging

pkg of 10 vials

technique(s)

microbiological culture: suitable

application(s)

food and beverages
microbiology

compatibility

for use with Listeria-Enrichment-Broth (Base)
for use with GranuCult® Buffered Listeria Enrichment Broth (base)

storage temp.

2-8°C

suitability

Listeria spp.

General description

Listeria selective enrichment supplement is a mixture of two antibiotics and a dye in lyophilized form. It inhibits the growth of accompanying bacteria for the selective enrichment of Listeria monocytogenes.

Application

Listeria selective enrichment supplement is an additive for the preparation of Listeria enrichment broth (Base) and Buffered Listeria enrichment broth.

Linkage

Replaces: 1117810017

Analysis Note

Supplement tested in Listeria-Enrichment-Broth (Base) acc. to FDA/IDF-FIL art. no. 1.11951.
Growth
- Listeria monocytogenes ATCC 19118:
good to very good
- Listeria monocytogenes ATCC 13932 (WDCM 00021): good to very good
- Listeria monocytogenes ATCC 35152 (WDCM 00109): good to very good
- Listeria innocua ATCC 33090 (WDCM 00017): good to very good
- Staphylococcus aureus ATCC 25923 (WDCM 00034): none to very good
- Escherichia coli ATCC 25922 (WDCM 00013): none
Incubation: 24 - 48 hrs.; 30 °C; aerobic.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Oral - Aquatic Chronic 2 - Carc. 2 - Eye Dam. 1 - Met. Corr. 1 - Muta. 2 - Repr. 1B - Skin Corr. 1B

Storage Class Code

6.1A - Combustible, acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service