Skip to Content
Merck

1.04354

SAFC

L-Histidine monohydrochloride monohydrate

EMPROVE® EXPERT, Ph. Eur., BP, ChP, JP

Pharma Manufacturing

Synonym(s):

His, L-α-Amino-β-(4-imidazolyl)propionic acid monohydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H9N3O2 · HCl · H2O
CAS Number:
Molecular Weight:
209.63
Beilstein:
4168261
MDL number:
UNSPSC Code:
12352107
EC Index Number:
211-438-9

Agency

BP
ChP
JP
Ph. Eur.

Quality Level

vapor pressure

<1 hPa ( 20 °C)

product line

EMPROVE® EXPERT

Assay

98.5-101.0% (alkalimetric)
99.0-101.0% (perchloric acid titration)

form

solid

pH

3.5-4.5 (20 °C, 100 g/L in H2O)

mp

254 °C (dec.) (lit.)

solubility

149.55 g/L

density

1.49 g/cm3 at 20 °C

application(s)

liquid formulation
parenterals
pharma/biopharma processes
pharmaceutical

storage temp.

2-30°C

SMILES string

O.Cl.N[C@@H](Cc1c[nH]cn1)C(O)=O

InChI

1S/C6H9N3O2.ClH.H2O/c7-5(6(10)11)1-4-2-8-3-9-4;;/h2-3,5H,1,7H2,(H,8,9)(H,10,11);1H;1H2/t5-;;/m0../s1

InChI key

CMXXUDSWGMGYLZ-XRIGFGBMSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Chemicals play an important role in the stabilization of a biologic drug during its manufacturing and formulation process – for instance, by preventing aggregation. We offer a wide range of high-quality stabilizers, buffers and salts to successfully purify and formulate your biomolecules. Specifically developed for high-risk applications, they are low in bioburden and endotoxins.
As part of our Emprove® Program, our raw materials are offered with extensive documentation facilitating compliance of your pharma and biopharma product, full supply chain transparency and risk mitigation. Our SAFC® portfolio of high-quality products for biopharmaceutical and pharmaceutical formulation and production withstands strict quality control procedures and is produced according to applicable cGMP guidelines.

Application

Histidine monohydrochloride is used in biopharmaceutical manufacturing as a buffer in formulation. It is often used along with histidine to prepare histidine buffer with the desired pH value.
Due to its low microbial and endotoxin limits, L-Histidine monohydrochloride monohydrate Emprove® Expert is especially suitable for high-risk applications.

Legal Information

Emprove is a registered trademark of Merck KGaA, Darmstadt, Germany
SAFC is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

This article outlines critical factors in long-term stability of proteins, and how different stabilizers protect again mechanical and thermal stress.

Related Content

The conjugation step in ADC production attaches a highly potent active pharmaceutical ingredient (HPAPI) to a mAb via a stable linker. Effective conjugation involves complex decisions, outlined on this page with services and products to assure success.

Learn more on mAb subcutaneous or intravenous formulation, and the relevant associated products, including excipients.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service