Skip to Content
Merck
All Photos(1)

Key Documents

1.10453

Millipore

APT agar

for microbiology

Synonym(s):

APT Agar (All-purpose agar cont. Tween®), All-purpose agar cont. Tween®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106213
NACRES:
NA.74

Quality Level

form

powder (dehydrated (DCM))

manufacturer/tradename

GranuCult®

technique(s)

microbiological culture: suitable

pH

6.5-6.9 (25 °C, 59.5 g/L in H2O, after autoclaving)

solubility

59.5 g/L

application(s)

food and beverages
microbiology

storage temp.

15-25°C

suitability

lactic acid bacteria

Analysis Note

Appearance (clearness): clear
Appearance (colour): brown
pH-value (25 °C): 6.5 - 6.9
Growth (Lactobacillus acidophilus ATCC 4356 (WDCM 00098)): good to very good
Growth (Lactobacillus casei ATCC 393 (WDCM 00100)): good to very good
Growth (Lactobacillus fermentum ATCC 9338): good to very good
Growth (Lactobacillus plantarum ATCC 14917 (WDCM 00104)): good to very good
Growth (Leuconostoc mesenteroides ATCC 9135 (WDCM 00108)): good to very good
Growth (Lactococcus lactis spp. lactis ATCC 19435 (WDCM 00016)): good to very good
Incubation: 48 hrs.; 35 °C; aerobic.

Footnote

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany
TWEEN is a registered trademark of Croda International PLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service