Skip to Content
Merck
All Photos(3)

Key Documents

1.01615

SAFC

Agar

extra pure

Pharma Manufacturing

Synonym(s):

Agar-agar, Gum agar

Sign Into View Organizational & Contract Pricing

Select a Size

1 KG
€208.00
25 KG
€3,293.00

€208.00


Estimated to ship on28 May 2025


Request a Bulk OrderRequest a Sample

Select a Size

Change View
1 KG
€208.00
25 KG
€3,293.00

About This Item

Linear Formula:
(C12H18O9)n
CAS Number:
MDL number:
UNSPSC Code:
12352201
EC Index Number:
232-658-1

€208.00


Estimated to ship on28 May 2025


Request a Bulk OrderRequest a Sample

Quality Level

form

solid

potency

11000 mg/kg LD50, oral (Rat)

pH

6.8 (20 °C, 100 g/L in H2O)

mp

90 °C

application(s)

pharmaceutical
semi-solid formulation
solid formulation

storage temp.

15-25°C

SMILES string

O1[C@@H]([C@@H]([C@@H]([C@H](C1OC2[C@H]3OCC2OC([C@H]3O)C)O)OC)O)CO

InChI

1S/C14H24O9/c1-5-8(16)13-11(7(21-5)4-20-13)23-14-10(18)12(19-2)9(17)6(3-15)22-14/h5-18H,3-4H2,1-2H3/t5?,6-,7?,8-,9+,10-,11?,12+,13+,14?/m1/s1

InChI key

GYYDPBCUIJTIBM-DYOGSRDZSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Finding the right excipient that matches your needs as well as regulatory demands can be a complicated challenge in formulation. With our application know-how and regulatory expertise, we support you in every step of development, scale-up, and production.
As part of our Emprove® Program, our raw materials are offered with extensive documentation facilitating compliance of your pharma and biopharma product, full supply chain transparency and risk mitigation. Our SAFC® portfolio of high-quality products for biopharmaceutical and pharmaceutical formulation and production withstands strict quality control procedures and is produced according to applicable cGMP guidelines.

Application

Agar is a commonly used ingredient in both food and pharmaceutical applications. In pharmaceutical application, agar has been used primarily for oral dosage forms.

Legal Information

Emprove is a registered trademark of Merck KGaA, Darmstadt, Germany
SAFC is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service