Skip to Content
Merck
All Photos(1)

Key Documents

1.01406

Millipore

GranuCult® prime Violet Red Bile Lactose (VBRL) agar

acc. ISO 4832 and FDA-BAM GranuCult , ISO 4832, FDA-BAM

Synonym(s):

Coliform detection media, Crystal Violet Neutral red Bile Lactose agar, Crystal-violet neutral red bile agar, Crystal-violet neutral red bile agar, VRB-agar (Crystal-violet neutral-red bile agar), VBRL agar, VRB agar, VRB-agar (Crystal-violet neutral-red bile agar), Violet Red Bile Lactose agar, Violet Red Bile agar

Sign Into View Organizational & Contract Pricing

Select a Size

500 G
€213.00
5 KG
€1,890.00

€213.00


Estimated to ship on25 May 2025


Request more information

Select a Size

Change View
500 G
€213.00
5 KG
€1,890.00

About This Item

UNSPSC Code:
41106213
NACRES:
NA.74

€213.00


Estimated to ship on25 May 2025


Request more information

Agency

APHA
FDA-BAM
ISO 4832

Quality Level

sterility

non-sterile

form

granular

packaging

pkg of 5 kg
pkg of 500 g

manufacturer/tradename

GranuCult® prime

storage condition

protect from light (Keep tightly closed and dry)

technique(s)

microbiological culture: suitable

color

pink

pH

7.3-7.5 (37 °C, 40 g/L in H2O, after autoclaving)

solubility

39.5 g/L

bulk density

670 kg/m3

application(s)

food and beverages
microbiology

storage temp.

15-25°C

suitability

coliforms

General description

GranuCult® prime VRB (Violet Red Bile Lactose) agar is used for the detection and colony counting of coliform bacteria. Crystal violet and bile salts inhibit the growth of the Gram-positive accompanying bacterial flora. Degradation of lactose to acid is indicated by the pH indicator neutral red, which changes its color to red, and by precipitation of bile acids. The enzymatic digest of animal tissue provides carbon and nitrogen sources for bacterial growth. Yeast extract primarily supplies the B-complex vitamins, while agar is the solidifying agent.

Application

GranuCult® prime Violet Red Bile Lactose (VBRL) agar is suitable for the detection and colony counting of coliform bacteria from food and animal feed, water, and other materials.

Features and Benefits

  • GranuCult® offers superior granulated culture media
  • Safe and sustainable due to reduced risks associated with fine dust and toxic substance inhalation, resulting in a safer work environment
  • Excellent wettability, solubility, and free-flowing properties
  • Convenient, with minimal component separation and clumping, even under warm or humid conditions
  • High batch-to-batch reproducibility
  • Prolonged shelf life of up to five years
  • High number of test strains exceeding all regulatory demands
  • Granulation technology allowS many supplements to be included, with no need to add these separately

Linkage

Replaces: 1014065007

Analysis Note

Appearance (clearness): clear
Appearance (colour): red
pH-value (25 °C): 7.2 - 7.6
Solidification behaviour (2 hrs., 45 °C): liquid
Growth promotion test in accordance with the current version of DIN EN ISO 11133.
Inoculum on reference medium (Escherichia coli ATCC 8739 (WDCM 00012)):
Colony count (Escherichia coli ATCC 8739 (WDCM 00012)):
Recovery on test medium (Escherichia coli ATCC 8739 (WDCM 00012)): ≥ 50 %
Colony colour (Escherichia coli ATCC 8739 (WDCM 00012)): purplish-red colonies with or without precipitate halo
Inoculum on reference medium (Escherichia coli ATCC 25922 (WDCM 00013)):
Colony count (Escherichia coli ATCC 25922 (WDCM 00013)):
Recovery on test medium (Escherichia coli ATCC 25922 (WDCM 00013)): ≥ 50 %
Colony colour (Escherichia coli ATCC 25922 (WDCM 00013)): purplish-red colonies with or without precipitate halo
Inoculum on reference medium (Enterobacter cloacae ATCC 13047 (WDCM 00083)):
Colony count (Enterobacter cloacae ATCC 13047 (WDCM 00083)):
Recovery on test medium (Enterobacter cloacae ATCC 13047 (WDCM 00083)): ≥ 50 %
Colony colour (Enterobacter cloacae ATCC 13047 (WDCM 00083)): purplish-red colonies with or without precipitate halo
Growth (Pseudomonas aeruginosa ATCC 27853 (WDCM 00025)): no limit
Colony colour (Pseudomonas aeruginosa ATCC 27853 (WDCM 00025)): colourless to beige colonies
Growth (Enterococcus faecalis ATCC 19433 (WDCM 00009)): total inhibition
Growth (Enterococcus faecalis ATCC 29212 (WDCM 00087)): total inhibition
Incubation: 24 ± 2 hours at 30 ± 1 °C
Reference media: Tryptic Soy Agar
A recovery rate of 50 % is equivalent to a productivity value of 0.5.
The indicated colony counts result from the sum of a triple determination.

Other Notes

  • The prepared medium is clear and red
  • If the medium is to be used immediately for the poured plate technique, cool it to 44-47 °C in a water bath before use
  • If the medium is used for the surface plating technique, there should be no visible moisture on the plates before use
  • The prepared plates can be stored at 2 °C to 8 °C in the dark and protected against evaporation for up to 5 days

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered®culture medi. depending on your needs. The designations basi. plu. or prime are added to indicate the quality control leve. from basic quality control to standard QC plus to prime for full regulatory compliance.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Water source comparison for microbiological media preparation highlights central purification vs. Milli-Q® IX system efficiency.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service