Skip to Content
Merck
All Photos(1)

Key Documents

1.09900

Supelco

Ammonium thiocyanate solution

c(NH₄SCN) = 0.1 mol/l (0.1 N), concentrate for 1000 mL titration solution, Titrisol®

Synonym(s):

Ammonium sulfocyanide

Sign Into View Organizational & Contract Pricing

Select a Size

1 AMP
€38.20

€38.20


Estimated to ship on30 May 2025



Select a Size

Change View
1 AMP
€38.20

About This Item

UNSPSC Code:
12352301
NACRES:
NA.21

€38.20


Estimated to ship on30 May 2025


Product Name

Ammonium thiocyanate solution, for 1000 ml, c(NH₄SCN) = 0.1 mol/l (0.1 N) Titrisol®

Quality Level

product line

Titrisol®

form

liquid

reaction suitability

reaction type: Redox Reactions

concentration

0.1 M

technique(s)

titration: suitable

pH

4.5 (20 °C in H2O)

density

1.03 g/cm3 at 20 °C

storage temp.

15-25°C

Application


  • 3D printed polycaprolactone/β-tricalcium phosphate/carbon nanotube composite: Investigated for its physical properties and biocompatibility, offering potential enhancements in 3D printing applications for tissue engineering (Wang et al., 2024).

  • Core decompression with β-tri-calcium phosphate grafts and platelet-rich plasma: Used for treating avascular necrosis of the femoral head, demonstrating improved clinical outcomes by enhancing bone regeneration and vascularization (Lyu et al., 2023).



Legal Information

Titrisol is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Supplementary Hazards

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service