Skip to Content
Merck
All Photos(1)

Key Documents

1.08017

Supelco

Hydrazine Test Kit, colorimetric

0.10-1.0 mg/L (N₂H₄), MQuant®

Synonym(s):

Hydrazine Visual Test Kit

Sign Into View Organizational & Contract Pricing

Select a Size

100 TESTS
€96.80

€96.80


Estimated to ship on30 May 2025


Request a Bulk OrderRequest more information

Select a Size

Change View
100 TESTS
€96.80

About This Item

UNSPSC Code:
41116116
NACRES:
NA.24

€96.80


Estimated to ship on30 May 2025


Request a Bulk OrderRequest more information

Product Name

Hydrazine Test, colorimetric, 0.10-1.0 mg/L (N2H4), for use with MCOLORTEST®

specific analyte(s)

hydrazine

Quality Level

measuring range

0.10-1.0 mg/L (N2H4)

compatibility

for use with MCOLORTEST®

detection method

colorimetric

storage temp.

15-25°C

General description

Method: colorimetric with color-matching vessel 0.10 - 0.25 - 0.5 - 1.0 mg/l NH MQuant®
This MQuant® Hydrazine test kit is designed for the colorimetric determination of hydrazine in boiler water and boiler feed water, utilizing a color-matching vessel. The colorimetric method involves a chemical reaction that produces a color change proportional to the concentration of hydrazine present in the sample. This reaction can be quantified using the item 1.08018 color-matching vessel for hydrazine testing. The test can be used in
· Boiler water
· Boiler feed water

Method
In acidic solution hydrazine reacts with 4-(dimethylamino)benzaldehyde to form a yellow compound. The hydrazine concentration is measured semiquantitatively by visual comparison of the color of the measurement solution with the color zones of a color-matching vessel.

Application


  • A novel dual-responsive colorimetric/fluorescent probe for the detection of N(2)H(4) and ClO(-) and its application in environmental analysis and bioimaging: This study introduces an innovative dual-responsive probe using the Hydrazine Test, highlighting its utility in environmental monitoring and biological imaging. This development offers substantial advancements in detecting and analyzing hydrazine and hypochlorite, critical for ensuring chemical safety and environmental health (Lv et al., 2024).

  • New 1,8-naphthalimide-based colorimetric fluorescent probe for specific detection of hydrazine and its multi-functional applications: This publication directly relates to Hydrazine Testing, detailing the development of a specific probe for hydrazine detection. The study showcases the critical role of Hydrazine Test kits in ensuring safe handling and accurate monitoring of hydrazine levels in various industrial and research applications (Lu et al., 2024).

Legal Information

MCOLORTEST is a registered trademark of Merck KGaA, Darmstadt, Germany
MQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

CorrosionExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Met. Corr. 1 - Skin Sens. 1

Storage Class Code

8B - Non-combustible, corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service