Skip to Content
Merck
All Photos(1)

Key Documents

1.04507

Millipore

myo-Inositol

for biochemistry

Synonym(s):

myo-Inositol, Hexahydroxycyclohexane, Cyclohexanehexol, meso-Inositol, Myo-Inosit

Sign Into View Organizational & Contract Pricing

Select a Size

50 G
€49.70
250 G
€123.00

€49.70


Estimated to ship on29 May 2025


Request a Bulk Order

Select a Size

Change View
50 G
€49.70
250 G
€123.00

About This Item

Empirical Formula (Hill Notation):
C6H12O6
CAS Number:
Molecular Weight:
180.16
MDL number:
UNSPSC Code:
12352104
EC Index Number:
201-781-2
NACRES:
NA.79

€49.70


Estimated to ship on29 May 2025


Request a Bulk Order

Quality Level

form

solid

pH

5-7 (20 °C, 100 g/L in H2O)

mp

225-227 °C

solubility

143 g/L

storage temp.

15-25°C

SMILES string

OC1C(C(C(C(C1O)O)O)O)O

InChI

1S/C6H12O6/c7-1-2(8)4(10)6(12)5(11)3(1)9/h1-12H

InChI key

CDAISMWEOUEBRE-UHFFFAOYSA-N

Analysis Note

Assay (HPLC, calc. on anhydrous substance): ≥ 99 %
Identity (IR-spectrum): passes test
Melting range (lower value): ≥ 224 °C
Melting range (upper value): ≤ 227 °C
Heavy metals (as Pb): ≤ 0.001 %
Water: ≤ 0.5 %

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service