Skip to Content
Merck
All Photos(2)

Key Documents

1.07735

Supelco

Silica gel

beads, desiccant ~ 2 - 5 mm

Sign Into View Organizational & Contract Pricing

Select a Size

1 KG
€65.20

€65.20


Estimated to ship on26 May 2025


Request a Bulk Order

Select a Size

Change View
1 KG
€65.20

About This Item

CAS Number:
MDL number:
UNSPSC Code:
41123003
EC Index Number:
231-545-4
NACRES:
NB.21

€65.20


Estimated to ship on26 May 2025


Request a Bulk Order

Quality Level

form

beads

potency

>5000 mg/kg LD50, skin (Rabbit)

surface area

700 m2/g

loss

≤2% loss on drying, 180°C

particle size

2-5 mm

pore size

2.0-2.5 nm

pH

5-8 (20 °C, 100 g/L in H2O, slurry)

bp

2230 °C

mp

>1600 °C

bulk density

750‑800 kg/m3

storage temp.

no temp limit

SMILES string

O=[Si]=O

InChI

1S/O2Si/c1-3-2

InChI key

VYPSYNLAJGMNEJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • Chromatographic Applications: Silica gel is widely used as a stationary phase in chromatographic techniques for separating various chemical compounds. Innovations in silica gel modification, such as pentabromobenzyl group-modified silica gel, have enhanced the separation efficiency for metabolites like nicotinamide. This application is critical for pharmaceutical analysis and metabolomics studies, providing high precision and accuracy in quantifying biologically relevant molecules (Ozaki et al., Analytical Biochemistry, 2022).

  • Environmental Pollutant Detection: Silica gel is employed in creating molecularly imprinted polymers (MIPs) that selectively recognize and bind environmental pollutants. A notable application involves embedding quantum dots in molecularly imprinted silica gel to detect aminoimidazo-azaarenes (AIAs) in food samples. This method offers high selectivity and sensitivity, making it a powerful tool for food safety and environmental monitoring (Qiao et al., Current Analytical Chemistry, 2021).

  • Heavy Metal Removal from Water: Silica gel doped with specific reagents is used for the adsorption and removal of heavy metals from water samples. Recent research highlights the use of sol-gel silica doped with 3-(2-naphthoyl)-1,1-dibutylselenourea for the efficient removal of Pb(II) and Zn(II) from aqueous solutions. This application is essential for environmental remediation and ensuring water safety, showcasing the versatility and effectiveness of silica gel in addressing environmental pollutants (Mehmood et al., International Journal of Environmental Analytical Chemistry, 2022).

Analysis Note

Bulk density: about 75
Loss on drying (180 °C): ≤ 2 %
Water absorption capacity (24 hrs.,80 % relative humidity): ≥ 27 %

Storage Class Code

13 - Non Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Seham S El-Din El-Hawary et al.
Antioxidants (Basel, Switzerland), 8(12) (2019-12-19)
The prevalence of hepatic diseases globally and in Egypt particularly necessitates an intensive search for natural hepatoprotective candidates. Despite the traditional use of Chrysophyllum oliviforme L. and C. cainito L. leaves in the treatment of certain ailments, evidence-based reports on

Questions

1–5 of 5 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  3. When do I know I need to replace the silica? What color does it change to?

    1 answer
    1. This product does not have moisture indicating properties. A good alternative would be product 1019721000, which changes color from brown to yellowish upon water uptake from the atmosphere.

      Helpful?

  4. what is the color of this reagent (brown, orange & blue)?

    1 answer
    1. The silica gel product does not include an indicator. The color of this product is not apart of the test specification for this product.

      Helpful?

  5. Is this silica gel only recommended for dissectors under vacuum?

    1 answer
    1. This product is typically used in desiccators, with or without a vacuum. The particle size is 2 - 5 mm which may be a factor in considering this product for other conditions or applications. See the link below for the Drying Agent product brochure.
      Other desiccants in other formats are listed:
      https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/product/documents/124/148/br3537en-ms.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service