Skip to Content
Merck
All Photos(2)

Key Documents

1.01905

Supelco

Silica gel

granules, desiccant ~ 0.2 - 1 mm

Sign Into View Organizational & Contract Pricing

Select a Size

1 KG
€73.20

€73.20


Estimated to ship on02 June 2025


Request a Bulk Order

Select a Size

Change View
1 KG
€73.20

About This Item

CAS Number:
MDL number:
UNSPSC Code:
41123003
EC Index Number:
231-545-4
NACRES:
NB.51

€73.20


Estimated to ship on02 June 2025


Request a Bulk Order

Quality Level

form

granular

potency

>5000 mg/kg LD50, skin (Rabbit)

surface area

700 m2/g

loss

≤2.0% loss on drying, 180°C

particle size

0.2-1 mm

pore size

2.0-2.5 nm

pH

7 (20 °C, 100 g/L in H2O, slurry)

bp

2230 °C

mp

>1600 °C

bulk density

200‑800 kg/m3

storage temp.

15-25°C

SMILES string

O=[Si]=O

InChI

1S/O2Si/c1-3-2

InChI key

VYPSYNLAJGMNEJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • Stability and Structure of Potentially Atmospherically Relevant Glycine Ammonium Bisulfate Clusters: This research examines the complex interactions and stability of atmospheric aerosols involving sulfuric acid, providing insights valuable for environmental scientists and chemists studying atmospheric chemistry and pollution mitigation strategies (Hariharan et al., 2024).

  • Nanostructured Niobium and Titanium Carbonitrides as Electrocatalyst Supports: Discusses the role of sulfuric acid in synthesizing nanostructured materials for energy applications, relevant for material scientists and researchers in renewable energy technologies (McLeod et al., 2024).

  • Synthesis and characterization of hydroxyapatite nanoparticles from calcium hydroxide fouled with gases evolved from the smokestack of the glass industry: Highlights the utility of sulfuric acid in the synthesis of nanoparticles for industrial applications, crucial for researchers in materials science and industrial manufacturing (Abdelmoaty and Mousa, 2024).

  • Nucleophilic hydrolysis enables HF-etched MXene kilofarad specific capacitance and excellent rate performance: Demonstrates the application of sulfuric acid in enhancing the performance of electrochemical storage devices, relevant for material scientists working on advanced battery technologies (Xu et al., 2024).


Analysis Note

Bulk density: 60 - 70
Loss on drying (180 °C): ≤ 2.0 %
Water absorption capacity (24 hrs.,80 % relative humidity): ≥ 27.0 %

Storage Class Code

13 - Non Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service