Skip to Content
Merck
All Photos(1)

Key Documents

1.07651

Millipore

Sucrose

suitable for microbiology

Synonym(s):

α-D-Glc-(1→2)-β-D-Fru, α-D-Glucopyranosyl β-D-fructofuranoside, β-D-Fructofuranosyl-α-D-glucopyranoside, D(+)-Saccharose, Sugar

Sign Into View Organizational & Contract Pricing

Select a Size

1 KG
€83.80
5 KG
€435.00
25 KG
€1,260.00

€83.80


Estimated to ship on29 May 2025


Request more information

Select a Size

Change View
1 KG
€83.80
5 KG
€435.00
25 KG
€1,260.00

About This Item

Empirical Formula (Hill Notation):
C12H22O11
CAS Number:
Molecular Weight:
342.30
Beilstein:
90825
MDL number:
UNSPSC Code:
41106212
EC Index Number:
200-334-9
NACRES:
NA.74

€83.80


Estimated to ship on29 May 2025


Request more information

form

solid

Quality Level

potency

29700 mg/kg LD50, oral (Rat)

shelf life

limited shelf life, expiry date on the label

packaging

pkg of 1 kg
pkg of 25 kg
pkg of 5 kg

pH

7 (20 °C, 100 g/L in H2O)

mp

185-187 °C (lit.)

bulk density

800‑950 kg/m3

application(s)

microbiology

storage temp.

15-25°C

SMILES string

OC[C@H]1O[C@H](O[C@]2(CO)O[C@H](CO)[C@@H](O)[C@@H]2O)[C@H](O)[C@@H](O)[C@@H]1O

InChI

1S/C12H22O11/c13-1-4-6(16)8(18)9(19)11(21-4)23-12(3-15)10(20)7(17)5(2-14)22-12/h4-11,13-20H,1-3H2/t4-,5-,6-,7-,8+,9-,10+,11-,12+/m1/s1

InChI key

CZMRCDWAGMRECN-UGDNZRGBSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Sucrose can be used as a carbon source in microbial growth media. It provides a source of energy and carbon for many bacteria and supports their growth. Sucrose is used to produce several types of media for the isolation and cultivation of various bacteria, fungi, and yeasts. Sucrose is commonly used in media for the identification and differentiation of various bacteria that grow on sucrose-containing media. Chromogenic agar media, such as MacConkey agar media with sucrose, are also commonly used in the identification of bacterial strains. Sucrose can also be used in specialized media for the isolation and culture of certain organisms, such as lactic acid bacteria, which are commonly used in the food industry. Additionally, sucrose can serve as a differential reagent in media formulations, such as in the TSI test (triple sugar iron test), which differentiates various Enterobacteriaceae bacteria based on their ability to ferment various types of sugars.

Application

Sucrose is commonly used as an ingredient in many microbial culture media for the growth and identification of bacteria, yeasts and molds from various pharmaceutical, environmental, food and beverage samples.

Analysis Note

Identity (IR-spectrum): passes test
Spec. rotation (α 20/D; 26 %; water): +66.2 - +66.8 °
Heavy metals (as Pb): ≤ 0.001 %
TLC-Test: passes test
Loss on Drying (105°C): ≤ 0.5 %
Suitability for microbiology: passes test

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sophie Dekoninck et al.
Cell, 181(3), 604-620 (2020-04-08)
During embryonic and postnatal development, organs and tissues grow steadily to achieve their final size at the end of puberty. However, little is known about the cellular dynamics that mediate postnatal growth. By combining in vivo clonal lineage tracing, proliferation kinetics

Questions

  1. Hi, can I please know the concentration (%, wt) of this sucrose?

    1 answer
    1. This is not a solution, thus concentration is not determined. The product is not tested for purity, nor is it tested for trace amounts of other sugars. However, the minimum specification for moisture content is 0.5 % or less.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service