Skip to Content
Merck
All Photos(1)

Key Documents

1.07615

Supelco

Rubidium chloride

for analysis EMSURE®

Synonym(s):

Rubidium chloride

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
€376.00

€376.00


Estimated to ship on29 May 2025


Request a Bulk Order

Select a Size

Change View
25 G
€376.00

About This Item

Empirical Formula (Hill Notation):
ClRb
CAS Number:
Molecular Weight:
120.92
MDL number:
UNSPSC Code:
12352302
EC Index Number:
232-240-9
NACRES:
NA.21
form:
solid

€376.00


Estimated to ship on29 May 2025


Request a Bulk Order

Quality Level

product line

EMSURE®

Assay

≥98.0% (silver nitrate titration)

form

solid

potency

4440 mg/kg LD50, oral (Rat)

pH

7 (20 °C, 50 g/L)

bp

1390 °C

mp

715 °C

density

2.76 g/cm3 at 20 °C

bulk density

1000‑1200 kg/m3

anion traces

sulfate (SO42-): ≤0.2%

cation traces

Al: ≤0.001%
Cu: ≤0.0003%
Fe: ≤0.0003%
Mg: ≤0.0005%
N: ≤0.003%
Pb: ≤0.003%
Zn: ≤0.0003%

storage temp.

2-30°C

SMILES string

[Rb+].[Cl-]

InChI

1S/ClH.Rb/h1H;/q;+1/p-1

InChI key

FGDZQCVHDSGLHJ-UHFFFAOYSA-M

Analysis Note

Assay (argentometric): ≥ 98.0 %
Identity: passes test
Sulfate (SO₄): ≤ 0.2 %
Total nitrogen (N): ≤ 0.003 %
Al (Aluminium): ≤ 0.001 %
Cu (Copper): ≤ 0.0003 %
Fe (Iron): ≤ 0.0003 %
Mg (Magnesium): ≤ 0.0005 %
Pb (Lead): ≤ 0.003 %
Zn (Zinc): ≤ 0.0003 %

Legal Information

EMSURE is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service