Skip to Content
Merck
All Photos(1)

Documents

1.07315

Millipore

SABOURAUD 2% glucose agar

for the cultivation of dermatophytes

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106213

Quality Level

form

medium granules (dehydrated (DCM))

technique(s)

microbiological culture: suitable

pH

5.5-5.7 (25 °C, 47 g/L in H2O, after autoclaving)

solubility

47 g/L

bulk density

660 kg/m3

application(s)

clinical
microbiology

storage temp.

15-25°C

suitability

molds
yeasts

General description

for the cultivation of dermatophytes

Analysis Note

Appearance (clearness)clear to almost clearAppearance (colour)yellowish-brownpH-value (25 °C)5.4 - 5.8Solidification behaviour (2 hrs., 45 °C)liquidGrowth (Trichophyton mentagrophytes ATCC 18748)good to very goodGrowth (Trichophyton rubrum ATCC 28188)fair to goodGrowth (Microsporum gallinae ATCC 12108)good to very goodGrowth (Trichophyton ajelloi ATCC 28454)fair to very goodGrowth (Microsporum canis ATCC 36299)good to very goodGrowth (Aspergillus brasiliensis (formerly A. niger) ATCC 16404 (WDCM 00053))good to very goodGrowth (Geotrichum candidum DSM 1240)good to very goodGrowth (Penicillium commune ATCC 10428)good to very goodGrowth (Candida albicans ATCC 10231 (WDCM 00054))good to very goodIncubation: 7 days; 28 °C; aerobic.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service