Skip to Content
Merck
All Photos(1)

Documents

1.05470

Millipore

GranuCult® prime SIM (Sulfite Indole Motility) Agar acc. ISO 15213

suitable for microbiology

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106213
NACRES:
NA.74

form

medium granules (dehydrated (DCM))

Quality Level

manufacturer/tradename

GranuCult®

technique(s)

microbe id | specific enzyme detection: suitable
microbiological culture: suitable

pH

7.2-7.4 (37 °C, 30 g/L in H2O, after autoclaving)

solubility

30 g/L

bulk density

580 kg/m3

application(s)

food and beverages
microbiology

storage temp.

15-25°C

suitability

Escherichia coli
coliforms
enterobacteriaceae

Analysis Note

Appearance (clearness): clear
Appearance (colour): yellowish-brown
pH-value (25 °C): 7.1 - 7.5
Growth (Escherichia coli ATCC 25922 (WDCM 00013)): good to very good
Growth (Escherichia coli ATCC 4351): good to very good
Growth (Klebsiella pneumoniae ATCC 13883 (WDCM 00097)): good to very good
Growth (Enterobacter cloacae ATCC 13047 (WDCM 00083)): good to very good
Growth (Salmonella typhimurium ATCC 14028 (WDCM 00031)): good to very good
Growth (Proteus hauseri ATCC 13315): good to very good
Blacking (Escherichia coli ATCC 25922 (WDCM 00013)): -
Blacking (Escherichia coli ATCC 4351): -
Blacking (Klebsiella pneumoniae ATCC 13883 (WDCM 00097)): -
Blacking (Enterobacter cloacae ATCC 13047 (WDCM 00083)): -
Blacking (Salmonella typhimurium ATCC 14028 (WDCM 00031)): +
Blacking (Proteus hauseri ATCC 13315): +
Indole formation (Escherichia coli ATCC 25922 (WDCM 00013)): +
Indole formation (Escherichia coli ATCC 4351): +
Indole formation (Klebsiella pneumoniae ATCC 13883 (WDCM 00097)): -
Indole formation (Enterobacter cloacae ATCC 13047 (WDCM 00083)): -
Indole formation (Salmonella typhimurium ATCC 14028 (WDCM 00031)): -
Indole formation (Proteus hauseri ATCC 13315): +
Motility (Escherichia coli ATCC 25922 (WDCM 00013)): +
Motility (Escherichia coli ATCC 4351): +
Motility (Klebsiella pneumoniae ATCC 13883 (WDCM 00097)): -
Motility (Enterobacter cloacae ATCC 13047 (WDCM 00083)): +
Motility (Salmonella typhimurium ATCC 14028 (WDCM 00031)): +
Motility (Proteus hauseri ATCC 13315): +
Incubation: 18-24 hrs.; 35 °C; aerobic.

Footnote

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service