Skip to Content
Merck
All Photos(1)

Documents

1.02244

Sigma-Aldrich

Casein

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.21

form

solid

solubility

20.1 g/L (experimental)

storage temp.

20-25°C

General description

Casein is a calcium phosphate-binding protein that constitutes up to 80% of the total milk proteins. In bovine milk, it exists in four types such as αS1, αS2, β, and κ. Casein can adapt its structure according to the changes in the environment and assemble in colloidal structures to form casein micelles. Casein micelles constitute about 10% of the volume in skim milk.

Analysis Note

Appearance:Almost white, fine or fine granular powder, almost odorless slightly yellowish or grey-yellowish.
N (Nitrogen):14 - 15 %
P (phosphorus):0.7 - 0.8 %
Fats:≤ 0.2 %
Free acid (as lactic acid):≤ 0.1 %
Sulfated ash (800 °C):≤ 1.5 %
Loss on Drying (105°C):≤ 6 %
Suitability as enzyme substrate:conforms internal standard

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Invited review: Understanding the behavior of caseins in milk concentrates
Milena C, et al.
Journal of Dairy Science, 4772-4782 (2019)
Arun Ghosh et al.
Biomacromolecules, 10(7), 1681-1688 (2009-05-14)
The development of advanced materials from biorenewable protein biopolymers requires the generation of more exogenous bonds to maintain the microstructure and durability in the final products. Casein is the main protein of milk, representing about 80% of the total protein.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service