Skip to Content
Merck
All Photos(1)

Key Documents

1.07040

Millipore

Petri-dish rack

For up to 12 Petri dishes

Synonym(s):

Petri dish holder

Sign Into View Organizational & Contract Pricing

Select a Size

1 UNIT
€68.10

€68.10


Estimated to ship on26 March 2025


Request more information

Select a Size

Change View
1 UNIT
€68.10

About This Item

UNSPSC Code:
41104308
NACRES:
NB.14

€68.10


Estimated to ship on26 March 2025


Request more information

Quality Level

feature

to hold 12 dish(es)

packaging

pkg of 1 unit

technique(s)

microbiological culture: suitable (RTU culture media)

application(s)

food and beverages

storage temp.

room temp

General description

The Petri dish rack is an accessory of the Anaerocult system. It can hold up to 12 Petri dishes and comes with an integrated holding clip for the Anaerotest strip.

Application

The Petri dish rack is used to conveniently place the Petri dishes into the Anaerobic jar.

Legal Information

ANAEROCULT is a trademark of Merck KGaA, Darmstadt, Germany
ANAEROTEST is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10-13 - German Storage Class 10 to 13


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service