Skip to Content
Merck
All Photos(1)

Key Documents

1.06587

Supelco

di-Sodium hydrogen phosphate solution

(buffer stock solution) 1/15 mol/l

Synonym(s):

Disodium Phosphate Solution

Sign Into View Organizational & Contract Pricing

Select a Size

1 L
€67.40

€67.40


Estimated to ship on30 May 2025


Request a Bulk Order

Select a Size

Change View
1 L
€67.40

About This Item

UNSPSC Code:
12161703
NACRES:
NA.24

€67.40


Estimated to ship on30 May 2025


Request a Bulk Order

grade

analytical standard

Quality Level

pH

9.2 (20 °C in H2O, ready-to-use solution)

density

1.01 g/cm3 at 20 °C

application(s)

environmental
food and beverages
general analytical
industrial qc
pharmaceutical

format

mixture

storage temp.

15-25°C

General description

The accuracy of pH measurement results is directly affected by the buffer reference materials used in calibration. We have a distinguished range of buffer solutions and substances for the exact calibration of your pH measurement equipment. Using Certipur® reference materials, you benefit from maximum accuracy, reliability and convenience and avoid costly repeat analyses.

Our wide range of pH calibration buffers offers ideal products and packaging for your individual need. All buffer solutions are traceable to NIST (National Institute of Standards and Technology, USA) and PTB (Physical Technical Institute, Germany) primary reference materials. Buffers solutions are offered in 1L PE bottle, measured at 20 °C or 25 °C.

Please visit ISO certificates and Site Quality Self-Assessments to access the current certificates of accreditation.

Download your certificate at http://www.sigma-aldrich.com to view certified values, including uncertainty and expiry date.

Analysis Note

concentration: 0.0664 - 0.0670 mol/l

Legal Information

CERTIPUR is a registered trademark of Merck KGaA, Darmstadt, Germany

recommended

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service