Skip to Content
Merck
All Photos(1)

Key Documents

1.05699

Supelco

di-Lithium tetraborate

for analysis

Synonym(s):

di-Lithium tetraborate

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
€71.60
1 KG
€477.00

€71.60


Estimated to ship on29 May 2025


Request a Bulk Order

Select a Size

Change View
100 G
€71.60
1 KG
€477.00

About This Item

Empirical Formula (Hill Notation):
B4Li2O7
CAS Number:
Molecular Weight:
169.12
MDL number:
UNSPSC Code:
12352302
EC Index Number:
234-514-3
NACRES:
NB.21
grade:
for analytical purposes
form:
solid
solubility:
141.2 g/L

€71.60


Estimated to ship on29 May 2025


Request a Bulk Order

grade

for analytical purposes

Quality Level

Assay

≥98.0% (acidimetric)

form

solid

pH

9.1 (20 °C, 100 g/L in H2O, slurry)

mp

930 °C

solubility

141.2 g/L

density

2.35 g/cm3 at 20 °C

bulk density

330 kg/m3

storage temp.

2-30°C

SMILES string

[B].[B].[B].[B].[Li].[Li].O.O.O.O.O.O.O

InChI

1S/4B.2Li.7H2O/h;;;;;;7*1H2

InChI key

DPYKRXYVXDYLEY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Linkage

Replaces: LX0366A-2; LX0366A

Analysis Note

Assay (acidimetric): ≥ 98.0 %
Chloride (Cl): ≤ 0.005 %
Phosphate (PO₄): ≤ 0.002 %
Sulfate (SO₄): ≤ 0.005 %
Heavy metals (as Pb): ≤ 0.005 %
Ca (Calcium): ≤ 0.005 %
Fe (Iron): ≤ 0.001 %
K (Potassium): ≤ 0.005 %
Mg (Magnesium): ≤ 0.001 %
Na (Sodium): ≤ 0.005 %

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Repr. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service