Skip to Content
Merck
All Photos(1)

Key Documents

1.05445

Supelco

Zinc iodide starch solution

for analysis

Synonym(s):

Zinc diiodide

Sign Into View Organizational & Contract Pricing

Select a Size

500 ML
€47.00

€47.00


Estimated to ship on28 May 2025


Request a Bulk Order

Select a Size

Change View
500 ML
€47.00

About This Item

UNSPSC Code:
12352401
NACRES:
NA.21

€47.00


Estimated to ship on28 May 2025


Request a Bulk Order

Quality Level

form

liquid

pH

6.2 (20 °C in H2O)

density

1.02 g/cm3 at 20 °C

storage temp.

15-25°C

Analysis Note

Suitability as indicator: conforms

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Protocols

Analytical Method: Iodine in table salt

Preparation of a standard solution for Sulfide

Preparation of a standard solution for Formaldehyde

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service